TaqPath BactoPure Microbial Detection Master Mix is optimized for rapid low-level microbial detection even in the presence of inhibitors. For clinical assay developers and infectious disease labs that need a rapid method for accurate low-level microbial detection on a variety of targets using complex samples, we offer TaqPath BactoPure master mix.
TaqPath BactoPure Microbial Detection Master Mix is optimized for rapid low-level microbial detection even in the presence of inhibitors. For clinical assay developers and infectious disease labs that need a rapid method for accurate low-level microbial detection on a variety of targets using complex samples, we offer TaqPath BactoPure master mix.
Unlike competitors, TaqPath BactoPure master mix offers:
- The lowest limit of detection across common microbial targets
- GPR conformity for clinical lab use
Benefits of TaqPath BactoPure Microbial Detection Master Mix include:
- Low-level detection of pan bacterial, fungal, mammalian, viral, and antibiotic resistance markers
- Tolerance of inhibitors typically found in diagnostic applications from purified samples and crude lysates. The unique proprietary formulation allows robust performance even in the presence of substances that can inhibit PCR, such as heparin, hematin, or humic acid, thus increasing confidence when working with a variety of complex samples.
- Wide dynamic range
- Optimized for multiplexing—available in two formulations (with and without ROX) that enable single- to four-target detection per reaction
TaqPath BactoPure master mix can be used for low-level of detection of any bacterial, viral, and fungal DNA, as well as antibiotic resistant markers up to 1 copy/uL. Empower your clinical labs with TaqPath BactoPure master mix to detect low levels of microbial DNA with confidence.
Resources
For Laboratory Use.