Maximize IC performance using optimized columns
Thermo Scientific products for ion chromatography are designed to keep your research moving forward seamlessly. Our innovative ion chromatography systems, automated eluent generation products, Thermo Scientific Dionex IC PEEK Viper Fittings and specialized standards and reagents help streamline your analyses and solve your chromatographic analysis challenges.
Ion chromatography systems
Our range of innovative ion chromatography (IC) instruments provide innovative technology and unparalleled robustness. Whether you handle just a few samples or a heavy workload, routine or challenging analyses, we have an ion exchange chromatography system for your performance and affordability requirements.
Eluent generation cartridges
Eluent generation (EG) cartridges electrolytically produce high-purity hydroxide, carbonate, and methanesulfonic acid eluents for isocratic and gradient runs in Reagent-Free IC (RFIC) systems. There is no manual eluent preparation required. For complex carbohydrate analysis, we offer a Dual Eluent Generation (Dual EGC) mode to generate highly reproducible, accurate KOH/KMSA eluent gradients.
IC PEEK Viper fittings
Create easy, consistent, low-dead-volume connections that maximize the performance of your Thermo Scientific Dionex ion chromatography system using IC PEEK Viper fittings. These fittings ensure that the ferrule is correctly positioned for increased efficiency and sharp chromatographic peaks and are available in convenient kits tailored specifically to your IC system.
Ion chromatography standards and reagents
To help simplify your ion chromatography analyses, we have developed all the high-purity reagents and standards you need. Our complete line of Thermo Scientific Dionex standards, reagents, and chemicals can help minimize time-consuming reagents and standards validation and keep your system operating at peak performance. Our selection of IC standards and reagents includes:
  • Anion standards
  • Cation standards
  • Eluent concentrates
  • Suppressor regenerant concentrates
  • Chemicals and reagents for transition metal analysis
For Research Use Only. Not for use in diagnostic procedures.