Anza™ Alkaline Phosphatase
Anza™ Alkaline Phosphatase
Invitrogen™
Anza™ Alkaline Phosphatase
Invitrogen™ Anza™ Alkaline Phosphatase is intended for use in the removal of 3' and 5' phosphate groups that remain afterRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
IVGN2204500 Reactions
IVGN22082000 Reactions
Catalog number IVGN2204
Price (USD)
236.00
Each
Add to cart
Quantity:
500 Reactions
Price (USD)
236.00
Each
Add to cart
Invitrogen™ Anza™ Alkaline Phosphatase is intended for use in the removal of 3' and 5' phosphate groups that remain after Anza™ restriction enzyme digestion, such as for the dephosphorylation of vectors prior to insert ligation and removal of 5′ phosphates prior to end-labeling. Since the 5′ phosphate group is required for DNA ligation, treatment of vectors with Anza Alkaline Phosphatase prevents self-ligation and re-circularization, resulting in decreased vector background when cloning.
Features of Anza Alkaline Phosphatase:
• Validated for use with all Anza restriction enzymes
• Dephosphorylates in 15 minutes at 37°C
• Heat inactivated after 5 minutes at 80°C
• Fully active in Anza Buffer
• Dephosphorylates all types of DNA ends (blunt, 5'- and 3'-overhangs)
Flexible protocol
Some restriction enzymes prohibit or reduce the dephosphorylation process when digestion and dephosphorylation are performed simultaneously. Therefore, Anza Alkaline Phosphatase may be used in one of three ways depending upon the restriction enzyme used:
One-Step Protocol: Simultaneous restriction enzyme digestion and dephosphorylation in Anza buffer
Heat Inactivation Two-Step Protocol: Digestion is carried out first, followed by heat inactivation (20 minutes at 80oC) then dephosphorylation
Column Purification Two-Step Protocol: Digestion is carried out first, followed by column purification then dephosphorylation
No need for enzyme removal
Anza Alkaline Phosphatase is completely inactivated after 5 minutes at 80°C, so enzyme removal via phenol/chloroform extraction or column purification is not required prior to subsequent ligation steps. If the One-Step protocol was used, heat the reaction mixture for 20 minutes at 80oC to inactivate the restriction enzyme as well as the alkaline phosphatase. Heat inactivation of the alkaline phosphatase prevents residual phosphatase from dephosphorylating the DNA fragments added to the reaction mixture for ligation.
Anza™ Alkaline Phosphatase is part of the Anza™ Restriction Enzyme Cloning System, unifying the traditional cloning processes. Other Anza DNA Modifying Enzymes include the Anza T4 DNA Ligase Master Mix, Anza T4 PNK Kit, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit.
For more information on the Anza system, visit www.thermofisher.com/anza.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10X Buffer
Product TypeAlkaline Phosphatase
Quantity500 Reactions
Shipping ConditionApproved for shipment on Wet or Dry Ice
EnzymeAlkaline Phosphatase
Product LineAnza™
Unit SizeEach
Contents & Storage
500 μL Anza Alkaline Phosphatase
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -5 to -30°C.