advanced-lab-refrigerators-278x195
Thermo Scientific TSG series general-purpose laboratory refrigerators are designed for the demanding requirements of daily use in research, clinical, and industrial laboratories storing noncritical products such as reagents, media, proteins, and other samples requiring a controlled environment of 2° to 10°C.
  • Green refrigerants and ENERGY STAR certified
  • Enhanced sample security
  • Quiet operation and and low heat output (btu/hr)
Browse our complete portfolio or contact one of our experts to help you make a selection.
Choose a compact general purpose lab refrigerator
TSG Countertop Refrigerator
TSG Glass Door Undercounter Refrigerator
TSG Solid Door Undercounter Refrigerator
ES Series Undercounter Refrigerator
Equipped with datalogging and download capabilities via USB port to support clinical record management requirements all while providing solid performance needed in healthcare, research and industrial applications.
Remote alarm supports requirements of research for monitoring the most rigorous and demanding storage needs, all while using less energy and putting almost no heat into the work environment.
Remote alarm supports requirements of research for monitoring the most rigorous and demanding storage needs, all while using less energy and putting almost no heat into the work environment.For routine sample protection, performance, security and quality are required, but where laboratory space is limited (EU only).
Request a quote
Capacity1.83 cu. ft. (51.8L)5.5 cu. ft. (156L)5.5 cu. ft. (156L)5.3 cu. ft. (151L)
Temperature Range2° to 8°C2° to 9°C2° to 9°C1° to 11°C
Doors1111
Shelves3333
Interior (DxWxH)12.2 x 14.3 x 18.4 in. (310 x 363 x 467mm)20.5 x 19.5 x 23 in. (520 x 495 x 584mm)20.5 x 19.5 x 23 in. (520 x 495 x 584mm)17.7 x 19.7 x 27.8 in. (450 x 500 x 705mm)
Exterior (DxWxH)22.1 x 20.5 x 18.4 in. (561 x 521 x 467mm)26.9 x 23.6 x 32 in. (684 x 600 x 813mm)26.9 x 23.6 x 32 in. (684 x 600 x 813mm)23.4 x 23.4 x 33.3 in. (595 x 595 x 845mm)
Shipping Weight83 lb. (38kg)145 lb. (66kg)145 lb. (66kg)143 lb. (65kg)
Choose an upright general purpose lab refrigerator
Looking for refrigerators for medical-related applications?
Visit our Upright Medical Refrigerators & Freezers page to browse and learn more about our high performance refrigerators designed with special temperature control, access and organizational features.
Single door general purpose lab refrigerators
TSG Series
12 cu. ft. Glass Door Refrigerator
23 cu. ft. Glass Door Refrigerator
23 cu. ft. Solid Door Refrigerator
27 cu. ft. Glass Door Refrigerator
27 cu. ft. Solid Door Refrigerator
 Designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
Request a quote
Capacity12 cu. ft. (339L)23 cu. ft. (651L)23 cu. ft. (651L)27 cu. ft. (764L)27 cu. ft. (764L)
Temperature Range2˚ to 10˚C2˚ to 10˚C2˚ to 10˚C2˚ to 10˚C2˚ to 10˚C
Doors11111
Shelves
44444
Interior (DxWxH)20.75 x 21 x 45.75 in. (527 x 533 x 1162mm)28.5 x 24 x 61.75 in. (724 x 610 x 1569mm)28.5 x 24 x 61.75 in. (724 x 610 x 1569mm)28 x 27 x 61.75 in. (724 x 686 x 1569mm)28.5 x 27 x 61.75 in. (724 x 686 x 1569mm)
Exterior (DxWxH)25 x 24.75 x 75.8 in. (635 x 629 x 1926mm)32 x 27.75 x 81 in. (813 x 705 x 2057mm)32 x 27.75 x 81 in. (813 x 705 x 2057mm)32 x 30.75 x 81 in. (813 x 781 x 2057mm)32 x 30.75 x 81 in. (813 x 781 x 2057mm)
Shipping Weight230 lb (105kg)392 lb (178kg)392 lb (178kg)386 lb (175kg)386 lb (175kg)
Double door general purpose lab refrigerators
TSG Series
45 cu. ft. Sliding Glass Door Refrigerator
49 cu. ft. Glass Door Refrigerator
49 cu. ft. Solid Door Refrigerator
 Designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
Request a quote
Capacity45 cu. ft. (1274L)49 cu. ft. (1387L)49 cu. ft. (1387L)
Temperature Range2˚ to 10˚C2˚ to 10˚C2˚ to 10˚C
Doors222
Shelves888
Interior (DxWxH)27 x 49 x 61.75 in. (686 x 1245 x 1569mm)28.5 x 49 x 61.75 in. (724 x 1245 x 1569mm)28.5 x 49 x 61.75 in. (724 x 1245 x 1569mm)
Exterior (DxWxH)32 x 52.7 x 81 in. (813 x 1339 x 2057mm)32 x 52.7 x 81 in. (813 x 1339 x 2057mm)32 x 52.7 x 81 in. (813 x 1339 x 2057mm)
Shipping Weight561 lb. (255kg)597 lb. (271kg)597 lb. (271kg)
Triple door general purpose lab refrigerators
72 cu. ft. Glass Door Refrigerator
 Designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
Request a quote
Capacity72 cu. ft. (2038L)
Temperature Range2° to 10°C
Doors3
Shelves12
Interior (DxWxH)28.5 x 72 x 61.75 in. (724 x 1829 x 1569mm)
Exterior (DxWxH)32 x 75.7 x 81 in. (813 x 1924 x 2057mm)
Shipping Weight733 lb. (332kg)
Which plug type do I need for my general purpose refrigerator?
 Standard Plug TypeVoltageHz
NEMA 5-15115–12060
NEMA 5-20115–12060
NEMA 6-15208–23060
Continental EU
23050
Related products
vaccine-storage-170x170
fms-freezer-170x170
deivce-link-hub-170x170
erlenmeyer-flasks-170x170
Related resources
Brochures:
TSX Series High-Performance Refrigerators
and Freezers
TSX Undercounter Refrigerators
ENERGY STAR Infographic