Open air shakers
Sample preparation is a vital step in your research, so choose our reliable open air laboratory shakers to focus more time on your objectives and less time managing your equipment. Open air benchtop shakers are ideal for protocols that use ambient temperatures such as staining, solubility studies, extractions and washing procedures. Open air shakers are suitable for use in incubators, warm rooms, environmental chambers or refrigerators, providing maximum versatility.
Choose shaker    Choose accessories
Reliability
reliable open air shaker
Our outstanding drive mechanisms provide continuous 24-hour operation and allow you to shake large and uneven loads, smoothly and uniformly.
Simplicity
Easy-to-use open air shaker
Easy-to-use controls are designed to precisely adjust temperature, speed and time. View all parameters simultaneously to prevent risk of error.
Flexibility
CO2-shaker-bottles-278x140
Suitable for many applications such as general cell culture, solubility studies, extraction procedures, general mixing, staining, destaining, washing procedures, and hybridizations.
What size open air shaker fits your needs?
 MaxQ CO2 Plus ShakerSolaris 2000
Shaker
Solaris 4000
Shaker
MaxQ 2508
Dual-Action Shaker
MaxQ 416 High Performance Shaker
 maxq co2 resistant shakermaxq 416 shaker
 External control box for ease-of-use without disrupting your samples.Combine a new ergonomic design with key feature enhancements that save you time and protect your processRuggedly built for demanding use and heavy loadsBack-and-forth or circular orbital motion. Ideal for bacteria growth, dissolution studies and staining of electrophoresis gels and chemical extractions.High performance triple counter-balanced drive handles sensitive cultures, plus a low-heat motor with CO2 resistant electronics.
 Learn moreLearn moreLearn moreLearn moreLearn more
Available universal platforms11 x 13 in.12 x 14in,
18 x 18 in, 18x24 in - each available in single or dual stack
18 x 30in,
24 x 36in - each available in single or dual stack
11 x 13 in.18 x 18 in. and
18 x 24in.
Speed30 to 300rpm
15 to 525 rpm
15 to 525 rpm15 to 525 rpm40 to 400 ±1rpm25 to 525 ±1rpm
Exterior dimensions
(H x W x D)
3.8 x 14.2 x 11.8 in.
(9.7 x 36 x 30cm)
5.5 x 14.3 x 18.5in.
(14.1 x 36.4 x 46.9cm)
7.0  x 22.6 x 25.5 in.
(17.7 x 57.3 x 64.8 cm)
7 x 13.5 x 17.4 in.
(17.8 x 34.3 x 44.2cm)
7.8 x 24.3 x 26 in.
(19.8 x 61.7 x 66cm)
Orbit.75 in. (1.9cm)1 in. (2.5cm)1 in. (2.5cm)0.4 in. (1.2cm) circular or fixed stroke angle1 in. (2.5cm)
Weight
capacity
13 lb. (6kg)42 lb. (19kg)42 lb. (19kg)15 lb. (6.8kg)71 lb. (32kg)
CO2 ResistantYesNoNoNoYes
AccessoriesPlatforms ›
Clamps ›
Racks & Misc ›
Platforms ›
Clamps ›
Racks & Misc ›
Platforms ›
Clamps ›
Racks & Misc ›
Platforms ›
Clamps ›
Racks & Misc ›
Platforms ›
Clamps ›
Racks & Misc ›
*Purchase of two platforms and adapter set required for stacking.
Want some help making a decision? Talk with one of our expert sales representatives.     Contact us
Optimize your open air shaker
open air shaker platforms
Universal platforms maximize flexibility by mixing different size labware on a single platform. View ›
open air shaker clamps
Various clamps maximize efficiency and productivity by mixing and matching vessel types with your universal platforms and customizing your shaker to fit your unique needs. View ›
open air shaker tube racks
Test tube racks and accessories outfit your shaker with holders that adjust to accommodate a wide variety of applications and objectives, and to keep conditions clean and safe. View ›
Universal platforms
Choose a platform that best fits your needs with flexibility for using a variety of labware on a single platform. Purchase clamps separately. Pick your ideal dimensions (L x W) below.
Choose a platform that best fits your needs with flexibility for using a variety of labware on a single platform. Purchase clamps separately. Pick your ideal dimensions (L x W) below.
Universal clamps
Mix and match to customize your universal platform for your unique needs. Clamps sold as individual units.
Mix and match to customize your universal platform for your unique needs. Clamps sold as individual units.
Test tube racks & accessories
The clamp alternative
shaker mat
Reliable, easy-to-use "Sticky Mats"
Thermo Scientific Adhesive Flask Mats, or "sticky mats," provide a flexible, convenient alternative for attaching a variety of vessels to any orbital shaker.
Top 3 best practices for using “sticky mats” on an orbital shaker
Order now
Five things to know about open air lab shakers
1. They operate in temperature ranges of 4°C to 40°C and humidity conditions from 20% to 80%, enabling them to be used in incubators, warm rooms, environmental chambers and refrigerators.
2. Open air orbital shakers can be found in multiple sizes with a variety of platforms as large as 36" x 24" and even stackable platforms in order to increase shaking capacity.
3. Open air lab shakers are typically the lowest cost options available, and there is no cost associated with compressors or heating mechanisms, but samples must be able to perform at ambient conditions.
4. Some manufacturers allow their shakers to be placed into environmental chambers, but always verify with the manufacture if this is allowed because if not it could void your warranty. Most that allow this require special reinforced shelving due to the weight and rotation of the shaker.
5. Some manufacturers allow their shakers to be placed into CO2 chambers for mammalian cell growth applications; these are treated to protect the electrical and mechanical components in the shaker from malfunction while exposed to high humidity and CO2. Again, it's important to verify with the manufacturer whether this is allowed and if there are special requirements such as reinforced shelving.
Related products