Notes
- Do not install this patch unless you have upgraded your GeneChip™ Scanner 3000 for high-resolution processing.
- If you plan to analyze array data generated from an instrument workstation or server that has been upgraded for high-resolution processing, you will need to apply this patch to your analysis workstation.
- The high-resolution scanning patch must be reapplied to GCOS workstations and server (if applicable) after installing all new library files.
For Research Use Only. Not for use in diagnostic procedures.