The Applied Biosystems StepOne and StepOnePlus Real-Time PCR Systems offer intuitive software, Fast (<40 min) and standard (<2 hr) runs, and are configured for PC-free or networked operation. These factory-calibrated systems make it easy to step up to high-performance qPCR. And now these systems support high resolution melt (HRM) software.
Request a quote Watch the video
Looking to commercialize? Contact our OEM services team today ›
StepOne System
The StepOne System is a remarkably simple real-time PCR system that enables precise quantitative real-time PCR results.
- Easy to use
- High-quality qPCR results
- Cost-effective, 3-color, 48-well instrument
- Designed with a user-friendly, powerful interface for researchers new to real-time PCR
StepOnePlus System
The StepOnePlus System includes additional performance features supporting the full range of Applied Biosystems TaqMan® Assays, while providing ease of use and a small footprint.
- Easy to use
- High-quality qPCR results
- 4-color, 96-well PCR instrument
- Innovative Applied Biosystems VeriFlex Block for convenient PCR optimization
Don’t forget to choose from our wide portfolio of quality assays, reagents, and plastics to run your experiments.
Keep your instrument in peak condition with our calibration kits and verification plates.
![]() | Web Links:
| |
Video: Fundamentals of Real-Time PCR |
Don’t forget to choose from our wide portfolio of quality assays, reagents, and plastics to run your experiments.
Keep your instrument in peak condition with our calibration kits and verification plates.
![]() | Web Links:
| |
Video: Fundamentals of Real-Time PCR |
For Research Use Only. Not for use in diagnostic procedures.