Automated nanoparticle analysis
Nanometer-sized particles are present in many materials, from food additives, high-performance metals to catalysts used for process optimization. Characterizing the composition, size and shape of nanoparticles is an important first step in better understanding the relationship between nanoparticles and their performance and quality. 
The Thermo Scientific Automated Particle Workflow (APW) streamlines chemical analysis workflows by providing an all-in-one software package that improves research and product development process for industrial, academic, and government labs.
APW is an automated solution that streamlines the workflow, from acquisition to automated processing. With APW, you can quickly acquire statistically relevant, high-resolution (nanoscale) results over a large area. APW’s seamless integration with Thermo Scientific Talos (S)TEM and Thermo Scientific Spectra (S)TEM instruments and Thermo Scientific Maps EDS Software makes it possible to obtain data with higher statistics (50 in total vs. >500/hour)
Imaging and analyzing nanoparticles in an easy way: An exploration
Automated Particle Size Analysis for TEM
Scanning transmission electron microscopes and software for nanoparticle analysis
Spectra Ultra - Scanning transmission electron microscope for imaging and spectroscopy of beam sensitive materials
The Thermo Scientific Spectra Ultra is an aberration-corrected S/TEM that offers an industry-leading level of characterization capabilities for material science and semiconductor applications at the highest resolution on various samples. With the new Spectra Ultra S/TEM, the accelerating voltage becomes an adjustable parameter, just like probe current, and the massive Ultra-X EDX system enables chemical characterization of materials too beam-sensitive for conventional EDX analysis.
The system is housed in a fully redesigned enclosure with a built-in on-screen display for convenient specimen loading and removal. The system is delivered on a platform offering unprecedented mechanical stability through passive and (optional) active vibration isolation.
Spectra 200 - High throughput TEM and STEM microscope for all material science applications
Thermo Scientific Spectra 200 (S)TEM is a high-throughput, aberration-corrected, (scanning) transmission electron microscope for all material science applications designed to offer an exceptional level of mechanical stability and high imaging quality through passive and (optional) active vibration isolation.
The system is housed in a fully re-designed enclosure with a built-in on-screen display for convenient specimen loading and removal. For the first time, full modularity and upgradeability is available between uncorrected and single-corrected configurations with variable heights, allowing maximum flexibility for different room configurations.
Spectra 300 - High resolution TEM and STEM microscope for all material science and semiconductor applications
Thermo Scientific Spectra 300 (S)TEM is a high resolution, aberration corrected, scanning transmission electron microscope for all material science applications. It sets a new benchmark that delivers an exceptional level of mechanical stability and high imaging quality through passive and (optional) active vibration isolation.
Housed in a redesigned enclosure the Spectra 300 (S)TEM has a built-in on-screen display for convenient specimen loading and removal. For the first time, full modularity and upgradeability can be offered between uncorrected and single-corrected configurations with variable heights, allowing maximum flexibility for different room configurations.  
Talos F200X TEM - TEM microscope for high resolution TEM and STEM with accurate chemical quantification
The Thermo Scientific Talos F200X STEM is a scanning transmission electron microscope that combines outstanding high-resolution STEM and TEM imaging with industry-leading energy dispersive X-ray spectroscopy (EDS) signal detection. 2D/3D chemical characterization with compositional mapping is performed by 4 in-column SDD Super-X detectors with exceptional cleanliness. The Talos F200X scanning transmission electron microscope allows for fast and precise EDS analysis in all dimensions, along with high-resolution TEM and STEM (HRTEM and HRSTEM) imaging with fast navigation for dynamic microscopy.
Talos F200S TEM - TEM microscope for high productivity, high resolution TEM and STEM characterization with chemical quantification
The Thermo Scientific Talos F200S (S)TEM is a (scanning) transmission electron microscope that combines outstanding high-resolution STEM and TEM imaging with industry-leading energy dispersive X-ray spectroscopy (EDS). The Talos F200S (S)TEM features great versatility and high throughput in STEM imaging and allows for precise EDS analysis and high-resolution TEM (HRTEM) for dynamic microscopy.
Maps Software - Correlative electron microscopy and cross-platform imaging automation software
Thermo Scientific Maps Software is an imaging and correlative workflow software suite compatible with the full line of Thermo Scientific SEM, DualBeam (FIB SEM) and TEM platforms.
Scientists and researchers rely increasingly on nanoscale observations to inform the latest advances in research and analysis. It has, however, become apparent that high-resolution observations lose much of their utility in the absence of the larger macroscopic context. Observations from multiple sources must be linked, providing the necessary multi-scale and multi-modal insight for truly valuable data.
Maps Software provides a powerful imaging workflow automation package within an easy-to-use and robust platform. With just a few clicks, you can collect impactful data while preserving the context of your observations.
Velox Software - STEM and TEM software supporting material imaging and analysis with optional package for EDS analysis
Thermo Scientific Velox Software combines comprehensive access to (scanning) transmission electron microscope (STEM and TEM) optics and detectors in order to provide you with excellent experimental control. Velox Software offers high reproducibility, yield, and support for quantitative STEM and TEM material analysis.
Velox Software offers unique packages for energy-dispersive X-ray spectroscopy (EDS, also called EDX) applications, in combination with the Thermo Scientific Super-X and Dual-X Detector Systems. A robust mapping engine combines multiple techniques optimized for transmission electron microscopy.  On the fly drift correction, recursive mapping, independent channel readout, and unique absorption correction with holder compensation are some of the techniques that ensure the best-in-class EDS spectrum image acquisition at high yields.
Avizo2D Software - 2D analysis software for materials characterization and quality control
Thermo Scientific Avizo2D Software provides advanced feature classification and statistics extraction capabilities on 2D images in a low learning curve environment.
Avizo2D Software allows materials scientists and quality control engineers to build and run image analysis ‘recipes’, combining deep learning models, pre-built workflows and powerful image processing tools.
For Research Use Only. Not for use in diagnostic procedures.