Search
This protocol outlines the steps for performing multiplexed staining of a tissue sample using a combination of purified primary antibodies and enzymatically mediated signal amplification assays. Two options for signal amplification are available: Invitrogen Aluora Spatial Amplification Kits and reagents or Invitrogen Alexa Fluor SuperBoost tyramide kits and reagents.
The use of signal amplification reagents is recommended for multiplexed imaging of tissue samples with low to medium expressed markers, especially when the signal obtained with antibody labeling alone is low. Aluora Spatial Amplification Kits can improve the signal-to-background ratio and boost signal. It is also beneficial for high abundance markers as it allows for reduced antibody usage.
For tissue samples and multiplexing multiple markers, we recommend utilizing the Aluora Spatial Amplification Kits and reagents. Tissues labeled with Aluora dyes are prepared for subsequent labeled-antibody or nuclei counterstaining steps, and can be imaged using any type of fluorescence microscope or spatial imager, including the Invitrogen EVOS S1000 Spatial Imaging System or Akoya PhenoImager.
It is important to note that this protocol assumes the availability of an automated slide stainer, such as the Leica BOND RX systems, to perform the required steps.
Leica BOND bulk solutions including:
The following negative and single-color controls are required for optimal spectral unmixing of the multiplex targets. Prepare your controls using the same workflow and equipment as multiplex sample. Please prepare each of the controls including:
| Single-color controls | Protocol |
|---|---|
| Single-color control: Make one for each primary antibody conjugate found in multiplex staining | Please follow the BOND protocol below. |
| Fill one titration kit container with one Aluora reagent. Replace other titration kit containers with wash buffers instead of dye. | |
| Nuclear counterstain only control | Add nuclear counterstain such as NucBlue or DAPI at the same concentration as was used in the multiplex sample. Replace other titration kit containers with wash buffers instead of stain. |
| Proceed to mounting step. | |
| Unstained tissue sample | No counter stain or any fluorophore conjugates |
| Proceed to mounting step |
We strongly recommend you optimize the primary antibody before doing the multiplex staining. Stain tissues with a serial dilution including 1:1,000 to 1:10,000 of the primary antibody conjugated to a fluorophore. If the target is highly expressed, some cases the dilution can be 1:50,000.
Create BOND system protocol for multiple rounds of staining from cycle 1 – 8. Follow BOND system manual to enter steps manually.
In the drop-down menu select pre-set preparation protocol. This should be based on your tissue type and needs. The step will include:
| Deparaffinization and rehydration | Temperature: 72°C | 30:00 | 150 μL |
Use the following settings for up to 8 staining cycles on the Leica BOND system.
| Step | Reagent | Temp | Incubation | Dispense | Staining cycle |
|---|---|---|---|---|---|
| 1 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 2 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 3 | Antibody Block | Ambient | 5:00 | 150 μL | 1 |
| 4 | Antibody 1 | Ambient | 30:00 | 150 μL | |
| 5 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 6 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 7 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 8 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 9 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 10 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 11 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 12 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 13 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 14 | Aluora 430 dye | Ambient | 10:00 | 150 μL | |
| 15 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 16 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 17 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 18 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 19 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 20 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 21 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 22 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 23 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 24 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 25 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 26 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 27 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 28 | Antibody Block | Ambient | 5:00 | 150 μL | 2 |
| 29 | Antibody 2 | Ambient | 30:00 | 150 μL | |
| 30 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 31 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 32 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 33 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 34 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 35 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 36 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 37 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 38 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 39 | Aluora 488 dye | Ambient | 10:00 | 150 μL | |
| 40 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 41 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 42 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 43 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 44 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 45 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 46 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 47 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 48 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 49 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 50 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 51 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 52 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 53 | Antibody Block | Ambient | 5:00 | 150 μL | 3 |
| 54 | Antibody 3 | Ambient | 30:00 | 150 μL | |
| 55 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 56 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 57 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 58 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 59 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 60 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 61 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 62 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 63 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 64 | Aluora 514 dye | Ambient | 10:00 | 150 μL | |
| 65 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 66 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 67 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 68 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 69 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 70 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 71 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 72 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 73 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 74 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 75 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 76 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 77 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 78 | Antibody Block | Ambient | 5:00 | 150 μL | 4 |
| 79 | Antibody 4 | Ambient | 30:00 | 150 μL | |
| 80 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 81 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 82 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 83 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 84 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 85 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 86 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 87 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 88 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 89 | Aluora 555 dye | Ambient | 10:00 | 150 μL | |
| 90 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 91 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 92 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 93 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 94 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 95 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 96 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 97 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 98 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 99 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 100 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 101 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 102 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 103 | Antibody Block | Ambient | 5:00 | 150 μL | 5 |
| 104 | Antibody 5 | Ambient | 30:00 | 150 μL | |
| 105 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 106 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 107 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 108 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 109 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 110 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 111 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 112 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 113 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 114 | Aluora 594 dye | Ambient | 10:00 | 150 μL | |
| 115 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 116 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 117 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 118 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 119 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 120 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 121 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 122 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 123 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 124 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 125 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 126 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 127 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 128 | Antibody Block | Ambient | 5:00 | 150 μL | 6 |
| 129 | Antibody 6 | Ambient | 30:00 | 150 μL | |
| 130 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 131 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 132 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 133 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 134 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 135 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 136 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 137 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 138 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 139 | Aluora 647 dye | Ambient | 10:00 | 150 μL | |
| 140 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 141 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 142 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 143 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 144 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 145 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 146 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 147 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 148 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 149 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 150 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 151 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 152 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 153 | Antibody Block | Ambient | 5:00 | 150 μL | 7 |
| 154 | Antibody 7 | Ambient | 30:00 | 150 μL | |
| 155 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 156 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 157 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 158 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 159 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 160 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 161 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 162 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 163 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 164 | Aluora 700 dye | Ambient | 10:00 | 150 μL | |
| 165 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 166 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 167 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 168 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 169 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 170 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 171 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 172 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 173 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 174 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 175 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 176 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 177 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 178 | Antibody Block | Ambient | 5:00 | 150 μL | 8 |
| 179 | Antibody 8 | Ambient | 30:00 | 150 μL | |
| 180 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 181 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 182 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 183 | HRP secondary Ab | Ambient | 10:00 | 150 μL | |
| 184 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 185 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 186 | *Bond Wash Solution | Ambient | 0:00 | Open | |
| 187 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 188 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 189 | Aluora 750 dye | Ambient | 10:00 | 150 μL | |
| 190 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 191 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 192 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 193 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 194 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 195 | *Bond ER Solution 1 | 95°C | 0:00 | 150 μL | |
| 196 | *Bond ER Solution 1 | 95°C | 20:00 | Intermediate | |
| 197 | *Bond ER Solution 1 | Ambient | 0:00 | 150 μL | |
| 198 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 199 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 200 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 201 | DAPI | Ambient | 5:00 | 150 μL | |
| 202 | *Bond Wash Solution | Ambient | 0:00 | 150 μL | |
| 203 | *Bond Wash Solution | Ambient | 1:00 | 150 μL | |
| 204 | *Bond Wash Solution | Ambient | 0:00 | 150 μL |
Dilute primary antibodies and make secondary antibody mix with equal volumes Poly-HRP-Goat Anti-Mouse IgG+ Poly-HRP-Goat Anti-Rabbit IgG. If using biotinylated primaries, include a biotin/strep-avidin blocking steps in that staining cycle and use the HRP-streptavidin reagent instead of poly-HRP antibody. Make up Aluora dye staining solution according to Aluora Spatial Amplification Kit user guide.
Run the programmed staining cycles. If additional support is needed, contact the Leica customer service.
Mount the coverslips using a mountant with antifade properties:
Analyze the tissue using a compatible imaging instrument.
PhenoImager is trademarked by Akoya Biosciences. BOND are trademarks of Leica Biosystems and its affiliates.
Invitrogen EVOS S1000 can be purchased in participating countries and regions including US, Canada, and Europe for 2024.
For Research Use Only. Not for use in diagnostic procedures.