Search
The IC installer automatically detects the current operating system and installs the appropriate IC version (IC 4.5 for Windows 2000 and IC 5.0 for Windows XP).
Note: Before downloading and installing IC 5.0, please review the software validation document for important compatibility information:
Windows 2000 and XP support and validation for software and instrumentation ›
Note: The GCS 3000 Low Resolution scanner (without the high resolution scanning patch) requires IC 3.1. Please contact your local Technical Support Center to obtain IC 3.1 or upgrade your scanner with the High-Resolution patch available below.
For Research Use Only. Not for use in diagnostic procedures.