Unleashing new potential for metabolomics, lipidomics, proteomics, and structural biology
Whether you are analyzing small molecules or large biomolecules, the Thermo Scientific Orbitrap Excedion Pro Hybrid Mass Spectrometer provides advanced performance and precision to help you achieve groundbreaking discoveries in your scientific analyses.
Featuring excellent sensitivity and intra scan dynamic range, the innovative Orbitrap Excedion Pro MS goes beyond the capabilities of traditional hybrid or Quadrupole Time-of-Flight (QTOF) MS technologies. Explore the full potential of your research with the cutting-edge technology of the Orbitrap Excedion Pro MS.
Highlights: Higher sensitivity with alternative fragmentation and new capabilities
Fast and sensitive alternative fragmentation
Electron Transfer Dissociation (EASY-ETD) option combined with HCD (Higher-Energy Collisional Dissociation) fragmentation delivers unique EThcD (Electron-Transfer/Higher-Energy Collisional Dissociation) fragmentation data on a hybrid benchtop mass spectrometer.
Lower limits of detection for a wider range of molecules
For increased ion flux and gentle transfer of ions for lower limits of detection for a wider range of molecules, the high-sensitivity atmospheric pressure interface combines high-capacity ion transfer tube and electrodynamic ion funnel, optimized for labile compounds and efficient ion transfer.
Superior acquisition rate and resolution
MS/MS acquisition rates of up to 70 Hz in the Orbitrap mass analyzer with maximum resolution of 480,000 (FWHM) at m/z 200.
Enhanced intra scan dynamic range
Enhanced dynamic range (eDR) mode enables five orders of magnitude intrascan dynamic range.
Sensitivity, specificity, and unbiased discovery combined
Hybrid-DIA combines the sensitivity and specificity of targeted experiments with the unbiased discovery of DIA-based shotgun proteomics in a single experiment.
Improved acquisition rate in TMTpro 32plex reagent experiments
Unique processing mode powered by Phased Spectrum Deconvolution Method (ΦSDM) improves the acquisition rate of TMTpro 32plex reagents experiments.
Comprehensive targeted compound coverage across large sample cohorts
Adaptive RT for dynamic adjustment of chromatographic monitoring windows ensures reproducible measurements with sensitivity and specificity for comprehensive coverage of targeted compounds across large sample cohorts.
Features of Orbitrap Excedion Pro Mass Spectrometer
With the Orbitrap Excedion Pro Mass Spectrometer, you don't just meet your goals; you exceed them! Embrace the future of mass spectrometry and unlock new possibilities in your research and development endeavors. Discover the power of innovation and exceed your expectations with the Orbitrap Excedion Pro MS.
Built on Orbitrap technology
Ultra-high-field Orbitrap mass analyzer with scan speed up to 70 Hz — Scan speed indicates the speed that the instrument acquires MS2 mass spectra per second.
Enhanced dynamic range (eDR) — Full MS mass range is dissected into two Orbitrap subscans, each performing multiplexed isolation and overall MS1 acquisition of equidistant or customizable m/z sub-ranges with efficient ion delivery to the Orbitrap analyzer. Stitching of two Orbitrap subscans is achieved automatically and fully embedded in the data structure. The number of windows within each Orbitrap subscan can be adjusted according to individual needs.
TurboTMT intelligent data acquisition mode (TMTpro 32plex) — Unique processing mode powered by Phased Spectrum Deconvolution Method (ΦSDM) specifically designed to improve the acquisition rate of Thermo Scientific TMTpro 32plex reagents experiments.
Hybrid-DIA — Combines the sensitivity and specificity of targeted proteomics experiments with the unbiased discovery of DIA-based shotgun proteomics in a single experiment.
Thermo Scientific OptaMax Plus Ion Source
Enhanced stability — The new heater and sprayer design significantly enhances heat transfer, ensuring exceptional electrospray stability across a broad temperature range.
User-friendly design — Experience enhanced ease of use with adjustable and reproducible movement to simplify your workflow.
Versatile flow rate compatibility — Supports a wide range of flow rates from 1 - 2,000 µL/min, providing flexibility for various applications.
Electrodynamic ion funnel (EDIF)
Optimized for labile compounds — Radio frequency (RF) device optimized for labile compounds efficiently capture ions as they leave the High-Capacity Transfer Tube (HCTT). Automatic tuning results in broad ion transmission curves with reduced ion losses, increasing sensitivity.
EASY-ETD option
Fast and sensitive alternative fragmentation Electron Transfer Dissociation (ETD) generated from fluoranthene radical anions. The townsend discharge provides extremely stable and robust ionization. Compact size, located entirely within the footprint of the instrument.
Fast and sensitive alternative fragmentation
ETD and EThcD fragmentation occur in the ion routing multipole followed by detection in the Orbitrap mass analyzer. The EThcD combination provides b- and y-type and c- and z-type fragment ions in a single Orbitrap spectrum.
Bent trap
Reduces noise by preventing neutrals and high-velocity clusters from entering the quadrupole mass filter using a double bent design geometry. Axial field along the length of the rods improves ion transfer robustness. To increase duty cycles up to 150%, a pre-accumulation of ions can be modulated with Automatic Gain Control (AGC).
Improve sustainability with less maintenance
To reduce carbon footprint and overall maintenance, Orbitrap Excedion Pro Mass Spectrometers come standard with a single rough dry pump instead of two oil pumps.
  • 30% less energy consumption
  • Reduced noise and heat production
Streamlined instrument control software
Software solutions across all next-generation Thermo Scientific platforms for ease of use through different systems. Get meaningful results faster with edition-specific application templates that are ready to use and built into the method editor.
Resources
Discover how you can unlock the full potential of your research with innovative features and capabilities.
Review precise specifications to determine the Orbitrap Excedion Pro Mass Spectrometer that meets your needs.
Discover how Hybrid DIA on the Orbitrap Excedion Pro mass spectrometer helps bridging global depth and targeted sensitivity for proteomic analysis.
See how Dr. Albert Heck and his team enhance sequence coverage for immunopeptidomics analysis with Orbitrap Excedion Pro MS and EThcD fragmentation.
Explore instrument technical details and request a quote
Service and support
Unity Lab Services provides world-class service solutions to support your instruments. Our comprehensive service portfolio was designed to meet the needs of your lab.
  • Instrument service plans
  • On-demand services
    • Compliance services
    • Preventive maintenance
    • Installation
  • Education
Managing services for your instrument has never been easier. Services Central is a centralized online platform that enables you and your colleagues to request service, view service request status and service history, check service plan or warranty coverage, access manuals, and more.
FAQs
The Orbitrap Excedion Pro Mass Spectrometer is designed for advanced research laboratories that require high-confidence molecular characterization across complex workflows. It is well suited for biopharma, academia, government, contract research (CRO), and other research laboratories performing proteomics, metabolomics, lipidomics, structural biology, small molecule, and biopharma applications.  
Its advanced fragmentation capabilities, high sensitivity, and high-resolution accurate-mass (HRAM) performance support laboratories that need to characterize challenging samples with confidence.
The Orbitrap Excedion Pro Mass Spectrometer expands the capabilities of the Orbitrap Excedion Mass Spectrometer for laboratories requiring more advanced biopharma, proteomics and multi-omics workflows.
It adds features and capabilities including the EASY-ETD option, Hybrid-DIA, TurboTMT intelligent data acquisition, scan speeds up to 70 Hz, and resolving power up to 480,000 at m/z 200. These features and capabilities support more comprehensive protein characterization, proteome and metabolome profiling, new quantitative capabilities, greater workflow flexibility, and improved performance for complex research applications.
Yes it is. With the Orbitrap Excedion Pro Mass Spectrometer, you can perform Hybrid DIA, a mass spectrometry (MS) acquisition strategy that combines DIA-based discovery proteomics with targeted peptide quantitation (PRM) in a single LC-MS analysis. The DIA part of the method provides broad, unbiased proteome coverage while targeted MS² scans (PRM) provide additional sensitivity and quantitative performance for predefined peptides of interest at the same time.
Instead of choosing between comprehensive discovery proteomics and sensitive targeted quantitation, hybrid DIA allows researchers to collect both types of information from the same sample in a single LC-MS analysis. This is particularly valuable when sample amount is limited or when an experiment contains known targets that must be measured reliably without sacrificing the opportunity to discover additional biological changes.
Yes. The Orbitrap Excedion Pro Mass Spectrometer can support both proteomics, metabolomics and even intact protein analysis on a single platform. This flexibility allows laboratories to perform metabolomics, lipidomics, proteomics, intact protein characterization, and other multi-omics workflows without requiring a separate mass spectrometer.
To improve laboratory productivity, the Orbitrap Excedion Pro Mass Spectrometer helps simplify method development, supporting faster data acquisition, and streamlining routine operation. Integrated instrument control software, application-specific method templates, advanced acquisition modes, and high-speed data collection enable laboratories to complete complex HRAM workflows more efficiently while maintaining high-quality analytical performance.
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only - Not For Diagnostic Procedures.