Search
Designed for today's fast-paced laboratories, the Mindray BS-480 and BA-800M chemistry analyzers are versatile workhorses offering mid to high volume throughput. The wide drugs of abuse testing menu serves the needs of clinical laboratories and criminal justice and forensics drug court programs. Thermo Fisher Scientific is the exclusive distribution partner of Mindray North America for these systems in the United States and Canada.
Mindray BS-480 Chemistry Analyzer
Mindray BA-800M Chemistry Analyzer
| Feature | BS-480 | BA-800M |
|---|---|---|
| Throughput | 400 Test Results/Hour | 800 Test Results/Hour |
| Per 5 hours instrument Operation | 500-1000 Test Results | 1000-2500 Test Results |
| Per 10-Drug Test Panel, 5 Hours Operation | 50-100 Samples/Day | 100-250 Samples |
| # Sample Positions | 90 Total; 60 are barcode readable | 440 Total |
| # Reagent Positions | 80 | 120 |
| Reaction Volume | 120 - 360 µL | 100 - 360 µL |
| Sample Volume | 1.5 - 45 µL, step by 0.1 µL | 1.5 - 35 µL, step by 0.1 µL |
| Reagent Volume | R1: 45 - 350 µL | |
| R2-R4: 10 - 350 µL | 15 - 300 µL, step by 0.5 µL | |
| Weight | 661 lbs. | 1430 lbs |
| Dimensions (W x D x H) | 45 in x 28 in x 45 in | 91 in x 40 in x 47 in |
Having greater than 25 years of experience, Thermo Fisher Scientific offers the most extensive drugs of abuse screening immunoassay test menu in the industry.
| Drug Tests | Specimen Validity Tests | |||
|---|---|---|---|---|
| Amphetamine | Cotinine | Heroin Metabolite (6-AM) | Mitragynine (Kratom)* | Creatinine Detect |
| Barbiturate | Ecstasy | Hydrocodone | Opiates | General Oxidant Detect |
| Benzodiazepine | Ecstasy Plus | LSD** | Oxycodone | Gravity Detect |
| Buprenorphine | Ethyl Alcohol | Methadone | Phencyclidine | pH Detect |
| Cannabinoids | Ethyl Glucuronide* | Methadone Metabolite | Propoxyphene* | |
| Cocaine | Fentanyl | Methaqualone | UR-144/XLR-11* | |
*For Criminal Justice and Forensic use only. **Available only on BS-480
| Part Number | Description |
|---|---|
| BA49E-PA00002 | BS-480 Chemistry Analyzer, 110V, w/o ISE, with Barcodes, Glass Cuvettes, Water Drainage Module |
| BA49E-PA00001 | BS-480 Chemistry Analyzer, 110V, with ISE, Barcodes, Glass Cuvettes, Water Drainage Module |
| Part Number | Description |
|---|---|
| BA81E-PA00002 | BA-800M module of analyzer (220V) with SDM and Barcode Readers, w/o ISE module (including II.2, and III.1) |
| BA81E-PA00003 | BA-800M module of analyzer (220V) with SDM, Barcode Readers, and with ISE module (including II.1, II.2, and III.1) |
Contact our Technical Support Team
Phone: 1-800-232-3342, Option 2
Email: techservice.mgc@thermofisher.com