Which Mouse T Cell Isolation Product is Right for You?
  Affordable & pure, flow compatible cells   Highest purity for molecular assays   Untouched—most physiological relevant cells
Isolation method Positive isolation   Depletion or positive isolation   Negative isolation
Starting samples Any sample (whole blood, spleen, lymph node)   Any sample (whole blood, spleen, lymph node)   Spleen, lymph node cells*
# cells processed 2 x 109 cells   1–2 x 109 cellls   1 x 109 cells
Purity Higher >90%   Higher >90%   High >85%
Yield Standard >55%   Highest >95%   High >70%
Viability >90%   >90%   >90%
Isolation time ~60min   ~30 min   ~50 min
Isolation temperature cold   cold   cold
Applications Cellular assays (cell culture, cell activation/proliferation, cytokine studies) Molecular: isolation of proteins & nucleic acids   Depletion of unwanted T cells or Positive isolation for molecular: isolation of proteins & nucleic acids   Cellular applications for physiological studies (functional studies, adoptive transfer, flow analysis or further sorting)
Bead-free target cells Yes (but antibody present)   No   No
Compatible with flow cytometry /flow sorting Yes   No   Yes
Shelf life from production date 2 years   3 years   3 years
Required materials Magnets   Magnets   Magnets
  Order Now   Order Now   Order Now
* Lyse the red cells prior to use to improve the purity
THIS IS FOR MOUSE RESEARCH - NOT FOR HUMAN