pSilencer™ 4.1-CMV puro
AM5775: pSilencer™ 4.1-CMV puro
Features:
Vector Size: 4781 bp
Promoter: CMV
Ampicillin resistance gene
Puromycin resistance gene (SV40 promoter, SV40 polyadenylation signal)
ColE1 origin of replication
Vector Map and Related Information
Features:
Vector Size: 4781 bp
Promoter: CMV
Ampicillin resistance gene
Puromycin resistance gene (SV40 promoter, SV40 polyadenylation signal)
ColE1 origin of replication
Vector Map and Related Information
We recommend the following primers for sequencing:
Forward: 5'-AGGCGATTAAGTTGGGTA-3'
Reverse: 5'-CGGTAGGCGTGTACGGTG-3'
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (55 base pairs) between the BamH I and Hind III sites.
Forward: 5'-AGGCGATTAAGTTGGGTA-3'
Reverse: 5'-CGGTAGGCGTGTACGGTG-3'
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (55 base pairs) between the BamH I and Hind III sites.
Sequence:
For Research Use Only. Not for use in diagnostic procedures.