AM5770: pSilencer™ 3.1-H1 neo
PROPRIETARY -- DO NOT DISCLOSE
Features:
Vector Size: 4285 bp
Promoter: Human H1
Ampicillin resistance gene
Neomycin resistance gene (SV40 early promoter, SV40 early polyadenylation signal)
ColE1 origin of replication
Vector Map and Related Information
PROPRIETARY -- DO NOT DISCLOSE
Features:
Vector Size: 4285 bp
Promoter: Human H1
Ampicillin resistance gene
Neomycin resistance gene (SV40 early promoter, SV40 early polyadenylation signal)
ColE1 origin of replication
Vector Map and Related Information
We recommend the following primers for sequencing:
M13F(-40): 5'-GTTTTCCCAGTCACGAC-3'
3.0rev: 5'-GAGTTAGCTCACTCATTAGGC-3'
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (66 base pairs) between the BamH I and Hind III sites.
M13F(-40): 5'-GTTTTCCCAGTCACGAC-3'
3.0rev: 5'-GAGTTAGCTCACTCATTAGGC-3'
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (66 base pairs) between the BamH I and Hind III sites.
Sequence: