UV-Vis spectroscopy instruments
Industry-tested analytical solutions
Meet your analytical challenges with our complete line of Thermo Scientific UV-Vis/Vis Spectrophotometers. Our proven designs and user-friendly software help you quantify, assess purity, and more. Experience accuracy, ease-of-use, and reliability in an affordable package.
Go from samples to answers quickly and easily with UV-Vis and Vis instruments designed to streamline measurements with consistent, high-quality results, time after time. Rely on industry-tested solutions for industrial quality control and the education, pharmaceutical, chemical, food and beverage, life science, and basic research communities.
Explore our Special Savings Offers!
* Promotions available in US and Canada only
UV-Vis and Vis instrument selection guide
Choose the instrument family that’s right for you
SPECTRONIC
Recommended for:
  • Education
Applications
  • Live display of ABS & %T 
  • Quantitative analysis (Beer's Law)
  • Fast Scanning OD600 cell growth analysis
Models
Sampling
  • Standard sample compartment holds both cuvettes and test tubes
  • Spill proof, washable sample compartment
Instrument features
  • USB for data storage and PC control (optional)
  • Built-in cuvette and test tube racks
Interface
  • Simple on-board controls with color graphical screen
Software
  • Full visible spectrum scan in only 15 seconds
  • Quantitative analysis with a factor or up to four standards
  • Easy to use interface
  • Software platform: VISIONlite 5 Software available for remote computer control
Printer
  • USB-connected thermal or PCL6 printer
Validation package
  • No
GENESYS
Recommended for:
  • Education/Advanced teaching labs
  • Industrial QA/QC
  • Research
  • Life Science
Applications
  • QA/QC
  • Reaction monitoring 
  • Colorimetric analysis
  • Life Science analysis
Models
Sampling
Sample compartment accommodates a wide range of accessories (options vary by model)
  • Single cell holders
  • Supports test tubes (GENESYS 30/40/50)
  • 4 and 8-cell changer options (GENESYS 140/150/180 & BioMate 160)
  • 8-cell changer included (GENESYS 180)
  • Sipper, Peltier
  • Fiber optic probe opens (GENESYS 150/180 and BioMate 160)
Instrument features
  • Small footprint
  • Low stray light optical system delivers exceptional linearity and reproducibility
  • Affordable price and low cost of ownership 
  • Xenon flash lamp guaranteed for up to 3 years of maintenance-free use (GENESYS 50/150/180 models, BioMate 160 models)
  • Tungsten Halogen lamp (GENESYS 30/40/140 models)
Interface
  • On-board control with high-resolution, touch screen
Software
  • Intuitive touch screen software with full UV-Vis or Visible spectral range
  • Comprehensive set of on-board applications including:
    • Live Display
    • Quantitative analysis
    • Scanning
    • Kinetics
    • Fixed wavelength measurements
  • Software platform: VISIONlite 5 Software available for remote computer control
Printer
  • Snap-on printer available
  • Print via USB, Ethernet or optional Wi-Fi*
* available for specific models only
Validation Package
  • No
Evolution
Recommended for:
  • Industrial QA/QC
  • Pharmaceutical QA/QC
  • Research
  • Life Science
Applications
  • QA/QC
  • Pharmaceutical analysis
  • Kinetics experiments
  • Protein and DNA assays
  • Current USP, EP, and JP compliance
  • 21 CFR part 11 compliance*
*with Insight Pro Security Software
Models
Sampling
  • Wide range of accessories to help measure any sample including film holders, auto samplers, fiber optic probes, integrating sphere, and reflectance accessories
  • SMART accessories minimize instrument set-up time and operation errors with cell changers and sipper
  • Single and 8-position water or Peltier thermostatted cell holders
Instrument features
  • Fixed and variable bandwidth options
  • Xenon flash lamp guaranteed for up to 3 years of maintenance-free use
  • Extensive range of accessory options to meet a wide array of sampling requirements
Interface
  • Full-featured control and analysis PC software
Software
  • Fully capable Insight Pro PC software
  • Automated calculations and analysis for solving complex quantitative analyses
  • Comply with current 2019 USP, EP, or JP requirements
  • Validate your system with Insight Pro Software includes methods to run IQ/OQ verification testing
  • Optional Insight Pro Security Software available to achieve 21 CFR part 11 compliance
  • Software platform: Insight Pro Software
Printer
  • Print directly to any Windows® compatible printer
Validation Package
  • Validator for Insght Pro Software
  • Insight Pro Security Software
NanoDrop
Recommended for:
  • Education
  • Life Science Research
  • Biotechnology QA/QC
  • Biopharma research and manufacturing
Applications
  • DNA & RNA quantification 
  • Protein quantification
  • Colorimetric Protein assays (BCA, Bradford, Pierce660 etc)
  • OD600 and custom methods
  • dsDNA and RNA fluorescence detection highly concentrated samples (Acclaro Pro)
  • Kinetics
Models
Sampling
  • Sample-retention system allows direct measurements from 1-2 μL of sample   
  • Optional cuvette position offers experimental flexibility 
  • Pre-programmed applications based on sample, measurement and assay type 
  • Large dynamic range requires no dilutions
Instrument features
  • Small footprint
  • NanoDrop Ultra with wide dynamic range (1-27,500ng/μL for dsDNA), no dilutions necessary Acclaro Sample Intelligence technology for sample contaminant information
  • No consumables,  low cost of ownership
  • Xenon flash lamp guarantees maintenance-free use
  • added cuvette option for  experimental flexibility
  • Added fluorescence option for increased sensitivity and specificity 
  • LED status indicator bar
Interface
  • NanoDrop Ultra: Standalone unit, touchscreen interface; NanoDrop Lite Plus with local control screen; PC control for other models
  • All PC models compatible with Windows® 10/11 Pro (64)
  • Microsoft OneDrive, Google OneDrive, and Thermo Fisher Cloud-based applications for data storage and sharing for NanoDrop Ultra
Software
  • Pre-programmed applications for life science (all models)
  • Standalone, NanoDrop Ultra touchscreen interface with data transfer to PC via USB, WiFi, Ethernet
  • NanoDrop One algorithms for contaminant analysis
  • Software platform: NanoDrop-model specific software
  • Optional Software for 21 CFR Part 11 compliance
  • Optional Acclaro Pro software for increased accuracy for highly concentrated samples
Printer
  • WiFi and USB printing available for NanoDrop Ultra; NanoDrop Lite Plus specific printer option; direct printing from PC for all other models
Validation Package
  • IQOQ packages*
  • Security Software*
*Available for NanoDrop Ultra/UltraC/Ultra FL/UltraC FL and NanoDrop Eight
SPECTRONIC 200 UV-Vis Spectrophotometer
Support your teaching goals with the rugged, simple-to-use Thermo Scientific SPECTRONIC 200 spectrophotometer, the standard for quick and easy routine measurements in instructional and analytical laboratories.
GENESYS UV-Vis Spectrophotometers
Built for quality control, academia, and routine measurements, Thermo Scientific GENESYS Spectrophotometers give you a range of intuitive solutions for routine analysis and specialized measurements.
Evolution UV-Vis Spectrophotometers
Discover the next breakthrough material with Thermo Scientific Evolution Spectrophotometers. Simplified workflows, advanced technology, and an array of accessories consistently deliver the high-quality results you depend on.
NanoDrop UV-Vis Spectrophotometers and Fluorometers
Thermo Scientific NanoDrop UV-Vis Microvolume Spectrophotometers and Fluorometers feature a pioneering sample-retention system that enables direct measurements of 1-2 μL samples without dilutions and pre-programmed life science methods.
UV-Vis Spectrophotometer accessories
Extend your spectrophotometer’s capabilities
It’s easy to enhance the capabilities of your UV-Vis and Vis spectrophotometers with the right accessories and software. Upgrade your instrument hardware with a broad range of accessories for single- and multi-cell sample holders, temperature control units, sipper systems, fiber optic probes, interface cables, replacement lamps, standards, and more. Add optional software to enable higher resolution spectra scans, peak pick, scan overlay, kinetics, automated workflow calculations, performance verifications, and regulatory compliance. Choose the combinations that are just right for your needs.
For Research Use Only. Not for use in diagnostic procedures.