Search
Accessories for operation of the Bigfoot Spectral Cell Sorter include interchangeable nozzles, QC beads, and sheath fluid. Choose among a wide selection of fluorophores and functional dyes to identify, sort, and analyze cell populations. Add a Sasquatch software upgrade to support advanced analysis for spectral flow cytometry. Spectral unmixing capability is not available for all configurations. Please review options with your sales representative.
With the Bigfoot Spectral Cell Sorter, you can choose from 70-, 100-, 120-, 150- and 200-µm ceramic nozzle tips with adjustable pressure settings. A swap tip wizard simplifies setup of the nozzle tips.
Bigfoot QC Beads, stored in their own vial in the multi-sample loader, are essential for daily QC of the Bigfoot Spectral Cell Sorter, while Bigfoot Sheath Fluid is used to focus the hydrodynamic stream.
Sasquatch Software (SQS) comes standard with all Bigfoot Spectral Cell Cytometers.
In addition, Spectral Sorting and Analysis for SQS is an optional software upgrade for spectral flow cytometry. The module provides spectral sorting capabilities using up to 55 fluorescence detectors for resolution of 30 or more colors.
For Research Use Only. Not for use in diagnostic procedures.