Comprehensive, extensible, and intelligent platform
Thermo Scientific Proteome Discoverer Software is the ultimate toolbox for protein research, offering customizable workflows tailored to the complexities of modern proteomics. It simplifies the identification and quantification of proteins in complex biological samples, supporting a wide range of workflows, including peptide and protein identification, post-translational modifications (PTM) analysis, isobaric mass tagging, and label-free quantitation (LFQ). It handles data from diverse acquisition methods, such as data-independent acquisition (DIA), data-dependent acquisition (DDA), and wide window acquisition (WWA), ensuring comprehensive coverage.
Advanced tools like INFERYS™ Rescoring and the CHIMERYS™ intelligent search algorithm by MSAID significantly increase identification rates, protein coverage, and the discovery of unique peptides and PTMs. The software enhances protein quantitation for isobaric mass tags and LFQ, supporting multiple database search algorithms and dissociation techniques. Whether analyzing glycopeptides, top-down proteomics, proteoforms, or MS-based structural biology like chemical cross-linking (XL-MS), Proteome Discoverer Software enables scientists to extract deeper biological insights from their data.
Discover how to transform proteomics mass spectrometry data into insights
Key benefits
Comprehensive tools for proteomic studies
With a wide range of database search options and the ability to combine the output from multiple algorithms, Proteome Discoverer Software increases confidence in results and decision making through cross-validation.
Complete sequence coverage and confident identification of post-translational modifications (PTMs) is realized with support for multiple dissociation techniques including collision-induced dissociation (CID), higher-energy collision-induced dissociation (HCD), proton transfer reaction (PTR), ultra violet photo dissociation (UVPD) and electron transfer dissociation (ETD) in tandem with built-for-purpose search engines.
Customizable data analysis workflows
Simplify data analysis with customizable workflows supporting multiple quantitative methods. Achieve experimental flexibility using TMT, TMTpro, or other isotopic labeling strategies, or opt for label-free quantitation techniques. Map quantitative data to biological information with study management and advanced algorithms for peak selection and peptide quantification, maximizing information extraction from proteomics studies.
Confidently and easily facilitate data comparison, exchange, and verification in compliance with data standards developed by the HUPO Proteomics Standards Initiative (PSI).
Workflow nodes
The workflow system is composed of nodes that can connect in linear or branched pathways. These are highly customizable and pre-defined for common workflows such as TMT, LFQ, PTM identification and cross-linked peptide ID.
Click image to enlarge
Analysis nodes
Spectra are streamed from the top of the workflow and pass through nodes that perform filtering, peptide ID, quantification, FDR calculation, PTM site localization, bioinformatic analysis, and more.
Click image to enlarge
Search engine options
The flexible workflows of Proteome Discoverer Software allow for search engines to be used independently, in series, or in parallel to significantly increase the number of peptide and protein identifications.
Click image to enlarge
Click image to enlarge
Click image to enlarge
Click image to enlarge
Click image to enlarge
Manage, automate, process and share proteomics data
The Thermo Scientific Ardia Platform brings together scientific data and results into a centralized location, enabling users to interface intuitively with data, set up automated processing, and easily share results with colleagues and collaborators. Power your data analysis with the platform that brings your data, instruments, and applications together to deliver the future of proteomics workflows.
Featured resources
High-throughput, large cohort proteomic analysis
Proteome coverage and faster throughput for single-cell samples
Automated high-throughput proteomic analysis of stored blood cells
Deeper proteome coverage and faster throughput
Order Proteome Discoverer Software
An intelligent approach to proteomics search
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.