Search
Increase lab versatility with the Digital Shaking Dry bath, which can be used as a dry bath, shaker, or both.
These products have a 2 year warranty and are CE marked. Package contains power cord with adapters (when necessary).
| Heating / Shaking Bath Specifications | |
|---|---|
| Temperature | Ambient +5° to 100°C Fluctuation at 80°C = 0.9 |
| Control | Feedback control PID |
| Display | Digital |
| Shaking speed | |
| Orbit size diameter | 2mm / 0.8 in. |
| Dimensions | 10.9 x 13.1 x 6.7 in. (276 x 334 x 170mm) |
| Required accessories | Full, half, or quarter blocks |
| Weight | 18.3 lbs (8.3kg) |
| Power | 120V (60Hz), 230V (50/60Hz) |
| Order now | 88880027 (US) 88880028 (CN, EU, UK) |