Search
The BacMam system uses a modified insect cell virus (baculovirus) as a vehicle to efficiently deliver and express genes in mammalian cells with minimum effort and toxicity. Invitrogen offers a variety of reagents and services utilizing the BacMam system to expand your research efforts.
Advantages of the BacMam system include:
Learn more about the workflow, transduction process, and how this system can be used in your laboratory in the BacMam Technology Overview.
BacMam reagents can be used in a variety of mammalian cell types. View the cell types that have been transduced using BacMam technology and learn how to optimize conditions for your cell type.
The BacMam system has been used to create fluorescent protein-based sub-cellular markers and biosensors, and to introduce genes for pharmacological targets and functional proteins.
For Research Use Only. Not for use in diagnostic procedures.