Search
Which human T cell isolation kit is right for your application and sample type? Dynabeads has kits optimized for specific sample types broadly for whole blood, mononuclear cells (MNC) and bone marrow, or for those focused directly on MNC.
| Highest purity for molecular assays | Affordable & pure, flow-compatible cells | Extreme purity & physiological quality | Untouched—most physiological relevant cells | ||||
|---|---|---|---|---|---|---|---|
| Isolation method | Depletion or positive isolation | Depletion or positive isolation | Positive isolation | Negative isolation | |||
| Starting samples | Any sample (whole blood, MNC, bone marrow) | Any sample (whole blood, MNC, bone marrow) | Any sample (whole blood, MNC, bone marrow) | MNC | |||
| # MNC processed (mL whole blood processed) | 1-2 x 109 MNC 200 mL whole blood | 1-2 x 109 MNC 200 mL whole blood | 2 x 109 MNC 80 mL whole blood | 1 x 109 cells | |||
| Purity | Highest 98-100% | Highest 98-100% | Higher 95-99% | High 90-95% | |||
| Yield | Highest 95-100% | Highest 95-100% | High 70-85% | Standard 50-70% | |||
| Viability | >95% | >95% | >95% | >95% | |||
| Isolation time | ~30 min | ~30 min | ~60 min | ~50 min | |||
| Isolation temp. | Cold | Cold | Cold | Cold | |||
| Applications | Depletion of unwanted CD2+ T cells or positive isolation for molecular applications, e.g., isolation of proteins & nucleic acids | Depletion of unwanted CD3+ T cells or positive isolation for molecular applications, e.g., isolation of proteins & nucleic acids | Cellular assays (cell culture, cell activation/proliferation, cytokine studies) Molecular isolation of proteins & nucleic acids | Cellular applications for physiological studies (functional studies, flow analysis, study of resting/activated cells) | |||
| Bead-free target cells | No | No | Yes (but antibody present) | Yes | |||
| Compatible with flow cytometry/sorting | No | No | Yes | Yes | |||
| Shelf life from production date | 3 years | 3 years | 2 years | 2 years | |||
| Required materials | Magnets | Magnets | Magnets | Magnets | |||
| Order now | Order now | Order now | Order now |