Search
Combining X-ray photoelectron spectroscopy and SEM imaging for a more complete understanding of your materials.
Obtaining a comprehensive understanding of a sample often requires analysis using multiple instruments. While scanning electron microscopy (SEM) imaging and energy dispersive X-ray spectroscopy (EDX) can provide composition information, they may not capture the surface chemistry critical for assessing material performance. Conversely, X-ray photoelectron spectroscopy (XPS) can reveal surface chemistry, but additional high-resolution imaging may be necessary to elucidate the intricate relationship between chemistry and structure and its impact on material behavior.
What if you could harness the combined powers of SEM and XPS?
With the Thermo Scientific CISA (Correlative Imaging and Surface Analysis) Workflow, you can seamlessly merge datasets from our XPS and SEM instruments, enabling deeper insights into your samples.
The CISA Workflow integrates XPS with SEM analysis, providing precise quantitative chemical composition data alongside structural information from microscope imagery. This integration is valuable for studying surface and interface reactions, which is crucial in material research across diverse areas such as batteries, polymers, catalysts, metals, coatings, and packaging.
The CISA Workflow utilizes point-of-interest images to establish connections between XPS and SEM data. This enables precise targeting of identical positions on each sample during system transitions. Additionally, dataset alignment can be effortlessly confirmed through images, ensuring that data from both systems was collected at the same position for enhanced accuracy.
The dynamic CISA Workflow offers reliable high performance and accuracy between two surface analysis techniques, giving you a competitive edge in research.
Our learning resources offer valuable information and insights on the principles, techniques, and applications of XPS and surface analysis.
For Research Use Only. Not for use in diagnostic procedures.