Search
Anion exchange chromatography is a form of ion exchange chromatography where a negatively charged biomolecule binds to a positively charged resin.
The Thermo Scientific POROS anion exchange (AEX) resin portfolio offers four unique chemistries. These strong and weak AEX resins possess distinctive selectivity in bind/elute as well as flow-through operation to produce high purity elution.
POROS anion exchange applications:
Resin | POROS XQ | POROS HQ | POROS PI50 | POROS D50 |
Type of AEX resin | Strong | Strong | Weak | Weak |
Surface chemistry | Fully quaternized amine | Quaternized polyethylenimine (mixed amine) | Polyethylenimine (mixed amine) | Dimethylaminopropy |
BSA binding capacity | >140 mg/mL | 75 mg/mL | 80 mg/mL | 90 mg/mL |
pH range | 1–14 | 1–14 | 1–9 | 1–9 |
Pore size (Angstrom) | 1,100 | 2,000 | 2,000 | 1,050 |
Figure 1. POROS resin design features. High performance resins, like POROS, can simplify the development process, balance all of the requirements and help develop more efficient and cost effective purification processes.
POROS AEX resins are designed for high-dynamic binding capacity over a wide range of pH and conductivity conditions, thereby increasing process development flexibility and manufacturing throughput. The 50 μm particle size provides outstanding resolution for unprecedented impurity clearance independent of scale and flow rate.
POROS AEX resins are 50 μm, rigid, polymeric, ion‑exchange chromatography resins that can be used for the chromatography of biomolecules including recombinant proteins, monoclonal antibodies, DNA, viruses, and peptides. The resin backbone consists of cross-linked poly[styrene divinylbenzene]. A polyhydroxyl surface coating enables nonspecific binding and different surface chemistries to provide four high-quality anion exchanger resins.
Process development scientists are looking for new tools that will clear even more impurities in a single step while maintaining high recoveries and process flexibility. Watch this video to learn how Thermo Scientific POROS Ion Exchange chromatography resins address purification challenges of today’s biomolecules.
Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
Explore digital resource
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.