Search
Symbol: Lr
Date of discovery: 1961
Name origin: Ernest O. Lawrence
Appearance: unknown
Discoverer: Albert Ghiorso, et al.
Obtained from: man-made
Melting point: unknown
Boiling point: unknown
Density[kg/m3]: unknown
Molar volume: unknown
Protons/Electrons: 103
Neutrons: 159
Shell structure: 2,8,18,32,32,9,2
Electron configuration: [Rn]5f147s27p1
Oxidation state: 3
Crystal structure: unknown
Lawrencium, a synthetic element, is made from californium and has no known uses. Little is known about this element, but its appearance is likely to be silvery-white or gray. Preliminary work on the element indicates that it behaves like actinides. Eleven isotopes of lawrencium have been synthesized and the longest half-life of the eleven was 216 minutes. Lawrencium was named after Ernest O. Lawrence, who invented the cyclotron, an instrument designed to accelerate beams of charged particles.
Products
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.