Search
Our low bleed, midpolarity cyano and phenyl polysiloxane Thermo Scientific™ columns offer the ideal robustness, thermal stability, and performance required for difficult separations, enabling improved limits of detection with enhanced resolution of low-level analytes.
Column | Recommended Application |
|---|---|
| TraceGOLD™ TG-35MS GC Columns | Columns with a higher phenyl content that provides useful elution order and retention time changes, superior inertness that ensures excellent peak shape and sensitivity, and a stability and performance profile ideal for separating active environmental compounds such as polycyclic aromatic hydrocarbons (PAHs) |
| TraceGOLD™ TG-35MS AMINE GC Columns | Columns that utilize a chemically altered tubing surface that reduces peak tailing and eliminates the need for column priming when analysing amines and other basic compounds |
| TraceGOLD™ TG-1301MS GC Columns | Highly inert, long-lived columns that provides low bleed, excellent reproducibility and column-to-column consistency even with sensitive detectors like ECDs and MS |
| TraceGOLD™ TG-624SilMS GC Columns | Highly inert columns with the ideal performance and stability profile for the analysis of volatile organics |
| TraceGOLD™ TG-1701MS GC Columns | Columns with a mix of cyano and phenyl groups for increased polarity and a different elution order relative to less polar columns, optimal for confirmation analysis |
| TraceGOLD™ TG-17MS GC Columns | Particularly suited for applications that require more polarity than a 5% phenyl column, ideal for confirmational analysis, with excellent inertness suitable for active compounds such as pesticides |
| TraceGOLD™ TG-17SilMS GC Columns | Optimize separation when analyzing active environmental compounds with this low-bleed GC column with superior inertness |
| TraceGOLD™ TG-225MS GC Columns | Low-bleed columns with high thermal stability and improved efficiency optimized for the analysis of volatile organics |
| TraceGOLD™ TG-200MS GC Columns | Columns with an exceptionally inert phase, selectivity, and elution order optimized for difficult separations and for compounds that phenyl and cyano phases cannot resolve |
| TRACE™ TR-35MS GC Columns | Low bleed 35% phenyl polysilphenylene-siloxane with exceptionally low surface activity |
| TRACE™ TR-1701 GC Columns | Low bleed columns that provide a good starting point for method development in a wide range of applications |
| TRACE™ TR-50MS GC Columns | Low-bleed columns particularly useful for applications requiring a higher temperature and more polarity than a 5% phenyl column, and with an inertness that results in minimal peak tailing and decreased breakdown of sensitive samples |
| TRACE™ TR-225 GC Columns | Robust 50% cyanopropylphenyl polysiloxane columns manufactured to minimize risk of damage from contaminated carrier gas |
| TRACE™ TR-V1 GC Columns | Low-bleed thick-film columns suitable for MS detection and the analysis of volatile analytes |
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.