Get the most powerful data collection and analysis software for your Applied Biosystems™ 3130, 3500, or 3730 Genetic Analyzer.
Powered by Windows™ 7, these new versions of software offer:
  • Improved algorithms for more accurate sequencing
  • Automatic import of SNP information from public domains
  • Alignment with widely used software requirements
  • New, smaller computer with faster dual-core processor
  • Enhanced security features
Using a 3500 Genetic Analyzer? Don’t get left behind!
Update your Data Collection Software to version 3.1.
The new version delivers all of the features above plus:
  • 96-sample polymer pouch minimizes polymer waste in lower-throughput labs and applications
  • 14-day stability for on-instrument consumables which minimizes consumable waste and possibly lowers the price per reaction
  • No consumable-based instrument hard stops, giving you control of how much and how long consumables are utilized
  • Use a single 36 -cm array for both sequencing and fragment analysis applications to help decrease sample processing times and driving laboratory efficiency
Learn more about Data Collection Software 3.1 ›
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.