Search
Regulations continue to be increasingly stringent and the quality of results is more important than ever. When your instrument, sample handling, and methodology is pushing the limits, your chromatography vials and seals can’t be the broken link in the chain. Ensuring quality in every part of your workflow, including vials and closures, is critical to success. See the difference in Thermo Scientific Certified Vials and Closures. They uniquely address your compliance by providing three increasingly stringent levels of certification:
All of our premier vials and closures (listed below) are Specification Certified, which means they go through a certification process to guarantee they meet physical specifications necessary for use with autosamplers.
Are you required to qualify the vials and closures for your validated application? Eliminate this resource consuming step and use Certified Kits. We did the work for you! No risk for extractables, no risk of losing valued sample by taking the wrong vial and closures, which might not be compatible with the sample or solvent.
Each lot of certified kits is tested by HPLC/UV and GC/MS against 15 critical parameters. Certificate of Conformance included in every pack. Kits are delivered shrink-wrapped to ensure product cleanliness.
Certified Vial Kits (Unassembled and Assembled) | |||||||
|---|---|---|---|---|---|---|---|
| Cat | Certified Convenience Kit (Unassembled) | Vial | Glass | Patched | Cap Color/ Catalog | Septum | Pack of |
| CERT5000-580W | Certified 9mm Wide Opening SureStop Vial, 2mL Vial: C5000-1W | Clear | Yes | Blue C5000-51B | Ivory PTFE/Red Rubber | 100 | |
| CERT5000-582W | Certified 9mm Wide Opening SureStop Vial, 2mL Vial: C5000-2W | Amber | Yes | Blue C5000-51B | Ivory PTFE/Red Rubber | 100 | |
| CERT5000-578W | Certified 9mm Wide Opening SureStop Vial, 2mL Vial: C5000-1W | Clear | Yes | Blue C5000-54B | Red PTFE/White Silicone | 100 | |
| CERT5000-575W | Certified 9mm Wide Opening SureStop Vial, 2mL Vial: C5000-2W | Amber | Yes | Blue C5000-54B | Red PTFE/White Silicone | 100 | |
| CERT5000-593W | Certified 9mm Wide Opening SureStop Vial, 2mL Vial: C5000-1W | Clear | Yes | Blue C5000-55B | Blue PTFE/White Silicone, Pre-slit | 100 | |
| CERT5000-576W | Certified 9mm Wide Opening SureStop Vial, 2mL Vial: C5000-2W | Amber | Yes | Blue C5000-55B | Blue PTFE/White Silicone, Pre-slit | 100 | |
| CERT5000-80 | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-1 | Clear | No | Blue C5000-51B | Ivory PTFE/Red Rubber | 100 | |
| CERT5000-80W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-1W | Clear | Yes | Blue C5000-51B | Ivory PTFE/Red Rubber | 100 | |
| CERT5000-82W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-2W | Amber | Yes | Blue C5000-51B | Ivory PTFE/Red Rubber | 100 | |
| CERT5000-92W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-1W | Clear | Yes | Blue C5000-54B | Red PTFE/White Silicone | 100 | |
| CERT5000-992 | Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL Vial: C4000-9 | Clear | No | Black C5000-64B | Bonded Red PTFE/White Silicone | 100 | |
| CERT5000-78W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-1W | Clear | Yes | Black C5000-64B | Bonded Red PTFE/White Silicone | 100 | |
| CERT5000-75W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-2W | Amber | Yes | Black C5000-64B | Bonded Red PTFE/White Silicone | 100 | |
| CERT5000-69LV | Certified 9mm Screw Vial, 350µL Fused Insert Vial: C4000-LV1W | Clear | Yes | Black C5000-64B | Bonded Red PTFE/White Silicone | 100 | |
| CERT5000-72LV | Certified 9mm Screw Vial, 350µL Fused Insert Vial: C4000-LV2W | Amber | Yes | Black C5000-64B | Bonded Red PTFE/White Silicone | 100 | |
| CERT4010-91 | Certified 10-425 Screw Vial, 2mL Vial: C4015-11W | Clear | Yes | Black C4010-68A | Bonded Red PTFE/White Silicone | 100 | |
| CERT4015-83 | Certified 13-425 Screw Vial, 4mL Vial: C4015-1 | Clear | No | Black C4015-67A | Bonded Red PTFE/White Silicone | 100 | |
| CERT5000-79 | Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL Vial: C4000-9 | Clear | No | Gray C5000-75C | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 | |
| CERT5000-93W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-1W | Clear | Yes | Gray C5000-75C | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 | |
| CERT5000-76W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-2W | Amber | Yes | Gray C5000-75C | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 | |
| CERT5000-993 | Certified 9mm Wide Opening Total Recovery Screw Vial, w/10µL Reservoir Vial: C4000-9TR | Clear | No | Gray C5000-75C | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 | |
| CERT4015-96 | Certified Shell Vial 1mL with SepCap | Clear | No | — | Natural Integral Molded Polyethylene | 200 | |
| CERT4011-89W | Convenience Kit, Certified Crimp Top Vial, 2mL Vial: C4011-1W | Clear | Yes | Silver | PTFE/Synthetic Red Rubber | 100 | |
| Assembled Certified Vial Kits | |||||||
| CERT5000-175W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-2W | Amber | Yes | Black C5000-64B | Bonded Red PTFE/White Silicone | 100 | |
| CERT5000-193W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-1W | Clear | Yes | Gray C5000-75C | Bonded Red PTFE/ White Silicone, Pre-slit | 100 | |
| CERT5000-176W | Certified 9mm Wide Opening Screw Vial, 2mL Vial: C4000-2W | Amber | Yes | Gray C5000-75C | Bonded Red PTFE/ White Silicone, Pre-slit | 100 | |
Is your ROI corrupted by changing columns too often, instrument contamination and/or expensive frequent MS detector maintenance activities? Thermo Scientific Mass Spectrometry (MS) Certified Vials undergo a proprietary cleaning process that greatly reduces the background particulates along with their potential effect on high sensitivity chromatography. Each lot of our premium range of certified products has been tested by LC/MS and GC/MS to ensure:
Download White Paper: MS Certified Vials: Pre-cleaned and certified vials for mass spectrometry
MS Certified Vial Convenience Kits (Unassembled) | ||||||
|---|---|---|---|---|---|---|
| Cat | MS Certified Convenience Kit(Unassembled) | Glass | Patched | Cap Color | Septum | Pack of |
| MSCERT5000-34W | 9mm SureStop Screw Vial, 2mL with AVCS Closure | Clear | Yes | Blue | Blue Silicone/PTFE | 100 |
| MSCERT5000-35W | 9mm SureStop Screw Vial, 2mL with AVCS Closure | Amber | Yes | Blue | Blue Silicone/PTFE | 100 |
| MSCERT5000-40W | 9mm SureStop Screw Vial, 2mL with AVCS Closure | Clear | Yes | Blue | Blue Silicone/PTFE, Pre-slit | 100 |
| MSCERT5000-41W | 9mm SureStop Screw Vial, 2mL with AVCS Closure | Amber | Yes | Blue | Blue Silicone/PTFE, Pre-slit | 100 |
| MSCERT5000-30LVW | 9mm 200μL Fused Insert Screw Vial | Clear | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-31LVW | 9mm 350μL Fused Insert Screw Vial | Clear | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-32 | 9mm Wide Opening 1.7mL High Recovery Screw Vial with 30μL Reservoir | Clear | No | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-33TR | 9mm Wide Opening 1.5mL Total Recovery Screw Vial with 10μL Reservoir | Clear | No | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-341W | 9mm Wide Opening Screw Vial, 2mL | Clear | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-351W | 9mm Wide Opening Screw Vial, 2mL | Amber | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-36LVW | 9mm 200μL Fused Insert Screw Vial | Clear | Yes | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-37LVW | 9mm 350μL Fused Insert Screw Vial | Clear | Yes | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-38 | 9mm Wide Opening 1.7 mL High Recovery Screw Vial with 30μL Reservoir | Clear | No | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-39TR | 9mm Wide Opening 1.5mL Total Recovery Screw Vial with 10μL Reservoir | Clear | No | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-401W | 9mm Wide Opening Screw Vial, 2mL | Clear | Yes | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-411W | 9mm Wide Opening Screw Vial, 2mL | Amber | Yes | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-S34W | 9mm Wide Opening Screw Vial, 2mL, silanized | Clear | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-S35W | 9mm Wide Opening Screw Vial, 2mL, silanized | Amber | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-S40W | 9mm Wide Opening Screw Vial, 2mL, silanized | Clear | Yes | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-S41W | 9mm Wide Opening Screw Vial, 2mL, silanized | Amber | Yes | Grey | Bonded Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT4011-74W | 11mm Snap Top Vial, 2mL | Clear | Yes | Red | Clear PTFE/Clear Silicone, Pre-slit | 100 |
| MSCERT5000-134W | 9mm Wide Opening Screw Vial, 2mL | Clear | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT5000-135W | 9mm Wide Opening Screw Vial, 2mL | Amber | Yes | Blue | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT4015-135W | 13-425 Screw Vial, 4mL | Clear | Yes | Black | Bonded Clear PTFE/Clear Silicone | 100 |
| MSCERT4015-136W | 13-425 Screw Vial, 4mL | Amber | Yes | Black | Bonded Clear PTFE/Clear Silicone | 100 |
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.