Search
Tecan's Infinite M1000/M1000 PRO and Infinite F500 have received the LanthaScreen Certified Plus designation and Tecan’s Infinite F200/200 PRO have received the LanthaScreen Certified designation from Life Technologies which ensures that these readers are validated to strict standards in instrument setup and assay performance.
These technical notes provide a method for verifying that a fluorescent plate reader is able to detect a change in time-resolved fluorescence energy transfer (TR-FRET) signal, confirming proper instrument set-up and a suitable response. The method is independent of any biological reaction or equilibrium and uses reagents that are on-hand for the LanthaScreen assay.
| Assay Technology | Adapta & LanthaScreen Europium | GeneBLAzer | LanthaScreen Terbium | Predictor | Z'-LYTE |
|---|---|---|---|---|---|
| Modality | TR-FRET | FRET (bottom) | TR-FRET | FP | FRET |
| Setup Guide | |||||
| Application Note | - | - | - | View | - |
| Poster | - | - | - | - | - |
| Excitation | 317/20 | 409/12 | 332/20 | 530/5 | 405/12 |
| Emission 1 | 620/12 | 460/12 | 490/10 | 585/20 | 445/12 |
| Emission 2 | 665/12 | 530/12 | 520/20 | 585/20 | 520/12 |
| Assay Technology | Adapta & LanthaScreen Europium | GeneBLAzer | LanthaScreen Terbium | Predictor | Z'-LYTE |
|---|---|---|---|---|---|
| Modality | TR-FRET | FRET (bottom) | TR-FRET | FP | FRET |
| Setup Guide | |||||
| Application Note | - | - | View | - | |
| Poster | - | - | - | - | - |
| Excitation Filter (12.5 mm) | 340/30 (PV002125) | 415/20 | 340/30 (PV002125) | 535/25 | 415/20 |
| Emission 1 Filter (12.5 mm) | 615/10 | 460/20 | 495/10 (PV003125) | 590/20 | 460/20 |
| Emission 2 Filter (12.5 mm) | 665/10 | 535/25 | 520/25 (PV003125) | 590/20 | 535/25 |
| Dichroic Mirror | 510 | NA | 50% or 510 | 560 | 50% |
| Assay Technology | Adapta & LanthaScreen Europium | GeneBLAzer | LanthaScreen Terbium | Predictor | Z'-LYTE |
|---|---|---|---|---|---|
| Modality | TR-FRET | FRET (bottom) | TR-FRET | FP | FRET |
| Setup Guide | Reader not tested | ||||
| Application Note | - | - | - | - | - |
| Poster | - | - | - | - | - |
| Excitation Filter (12.5 mm) | 340/30 (PV002125) | 409/20 | 340/30 (PV002125) | NA | 415/20 |
| Emission 1 Filter (12.5 mm) | 615/10 | 460/20 | 495/10 (PV003125) | NA | 460/20 |
| Emission 2 Filter (12.5 mm) | 665/10 | 525/25 | 520/25 (PV003125) | NA | 535/25 |
| Dichroic Mirror | 510 | NA | 50% or 510 | NA | 50% |
| Assay Technology | Adapta & LanthaScreen Europium | GeneBLAzer | LanthaScreen Terbium | Predictor | Z'-LYTE |
|---|---|---|---|---|---|
| Modality | TR-FRET | FRET (bottom) | TR-FRET | FP | FRET |
| Setup Guide | |||||
| Application Note | - | - | - | - | - |
| Poster | - | - | View | - | - |
| Excitation | 317/20 | 409/12 | 332/20 | 530/20 | 405/12 |
| Emission 1 | 620/12 | 460/12 | 485/20 | 585/20 | 445/12 |
| Emission 2 | 665/12 | 530/12 | 515/20 | 585/20 | 520/12 |
* Filters (diameter) or optics shown as center wavelength/bandwidth (nm) & dichroic mirrors shown as cutoff wavelength (nm); Contact Tecan for information on ordering filters & dichroic mirrors suitable with the microplate readers listed above
** Some compatible filter sets are available for purchase from Life Technologies. Click on selected filter sets for more information
Infinite is a registered trademark and Safire2 is a trademark of Tecan Group Ltd.
For Research Use Only. Not for use in diagnostic procedures.