Search
Choose from a variety of innovative technologies for companion animal genomic testing. From predesigned and validated content to custom solutions, our flexible technologies are designed to provide rapid and reliable results to responsible breeders, veterinarians and researchers. Our solutions include microarrays, targeted genotyping-by-sequencing (GBS) solutions, short tandem repeat (STR) parentage kits, and Applied Biosystems Custom TaqMan SNP Genotyping Assays.
Custom marker panels allowbreeders, veterinarians and researchers to target the most informative and important parentage, traits and defect markers for their breeds and varieties. Use in conjunction with our Applied Biosystems AgriSeq genotyping-by-sequencing (GBS) solutions to help deliver unambiguous sequence-based genotyping results.
| Quantity | Cat. No. | |
| Custom Equine Genetic Traits and Parentage Panel | Varies | Contact sales representative › |
Custom marker panels allowbreeders, veterinarians and researchers to target the most informative and important parentage, traits and defect markers for their breeds and varieties. Use in conjunction with our Applied Biosystems AgriSeq genotyping-by-sequencing (GBS) solutions to help deliver unambiguous sequence-based genotyping results.
Our array-based genotyping solutions are ideal for the identification, validation and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays or customize an array specifically for your study.
Product | # SNPs | Array Format | Cat. No. | Price |
| Axiom Equine Genotyping Array | 670,796 | 96 array | 550583 | Contact a sales representative › |
| Axiom Equine Plus Genotyping Array | 670,800 | 96-array | 550624 | Contact a sales representative › |
Product | # SNPs | Array Format | Cat. No. | Price |
| Axiom Rabbit Genotyping Array | 200,000 | 96 array | 550669 | Contact a sales representative › |
Microsatellites, also referred to as short tandem repeats (STRs), are commonly used for parentage testing, identification, and forensics due to their informativeness and Mendelian inheritance. Our parentage testing kits provide all of the necessary reagents for simultaneous amplification of multiple microsatellite loci in a single multiplex PCR reaction.
These assays provide a highly flexible technology for detection of polymorphisms within any genome. Create your own assays by submitting target sequences to our secure assay design pipeline using the Applied Biosystems Custom TaqMan Assay Design Tool. This pipeline has successfully generated millions of assay designs by utilizing heuristic rules deduced from both manufacturing and assay performance data. TaqMan Assays can be run on any of our real-time PCR instruments or used as an endpoint assay on other third-party systems, including the Array Tape platform.
For Research Use Only. Not for use in diagnostic procedures.