Search Thermo Fisher Scientific
Thermo Scientific Cytomat Storage Series provides robot-friendly random access storage of up to 1512 standard microplates and can be easily integrated into any automated system for multiple applications. The series accommodates a range of temperatures (Storage: 4°C to 25°C) and humidities, as well as gaseous atmospheres. Cooled storage systems can ensure a dry climate, via inert gas, minimizing absorption of water by dimethylsulfoxide (DMSO).
Incubation/storage for cell culture applications 4 to 25°C, Humidity up to 90% r.H., ContraCon decontamination routine.
Fastest, mid-capacity incubation and storage solution for up to 105 (126) standard microplates with dependable sample protection, a temperature range of 4°C to 25°C, and controllable humidity.
Fastest, high-capacity incubation and storage solution for up to 210 standard microplates with dependable sample protection, a temperature range of 4°C to 25°C, and controllable humidity.
Two Cytomat 24 C425 incubators combined into one for a capacity of up to 1008 standard microplates, a a temperature range of +4°C to 25°C, and humidity to 90% r.H.
The Cytomat system delivers the microplate to a single point, offering “easy-teach” integration for any automated system. A single loading and unloading point – the transfer station – makes teaching simple and cuts down the programming time, making for an extremely robot-friendly design.
These transfer stations offer a versatile placement of the Cytomat in the lab.
This standard transfer station supplies the microplate in portrait format. A 90° turnable version is available for landscape presentation.
This linear transfer station is available in different lengths (300mm, 400mm, 500mm, and customized lengths) for extended reach and delivers the microplate in portrait format. Additional options are available for landscape presentation and lateral movement.
This swap station accelerates your lab automation handling system by providing two plate positions.
This plate lift is stepper motor driven (SMD) and offers microplate movement to a predefined height within a 500 mm stroke.
The right options to match your application and automation needs with internal barcode readers, stackers, copper chambers and more.
The barcode reader is located on the plate handler, allowing retrieval of information on every position, even when manually loaded.
The stainless steel, autoclavable Cytomat Stacker accommodates all standard microplates. Dedicated pitch sizes are available to optimize capacity for different types of microplates.
Solid pure antimicrobial copper chambers are proven to be effective against contamination.
Many more options available
The Cytomat system delivers the microplate to a single point, offering “easy-teach” integration for any automated system. A single loading and unloading point – the transfer station – makes teaching simple and cuts down the programming time, making for an extremely robot-friendly design.
These transfer stations offer a versatile placement of the Cytomat in the lab.
This standard transfer station supplies the microplate in portrait format. A 90° turnable version is available for landscape presentation.
This linear transfer station is available in different lengths (300mm, 400mm, 500mm, and customized lengths) for extended reach and delivers the microplate in portrait format. Additional options are available for landscape presentation and lateral movement.
This swap station accelerates your lab automation handling system by providing two plate positions.
This plate lift is stepper motor driven (SMD) and offers microplate movement to a predefined height within a 500 mm stroke.
The right options to match your application and automation needs with internal barcode readers, stackers, copper chambers and more.
The barcode reader is located on the plate handler, allowing retrieval of information on every position, even when manually loaded.
The stainless steel, autoclavable Cytomat Stacker accommodates all standard microplates. Dedicated pitch sizes are available to optimize capacity for different types of microplates.
Solid pure antimicrobial copper chambers are proven to be effective against contamination.
Many more options available
The Cytomat 10 automated incubator series combines superior environmental control, fast access times and versatility, making it the ideal choice for cell-based workflows requiring larger capacity incubation.