Search
Join the conversation
Primary XPS region: S2p
Overlapping regions: Si2s plasmon, Se3p
Binding energies of common chemical states:
| Chemical state | Binding energy S2p3/2 |
|---|---|
| Metal sulfide | ~161.5 eV |
| Thiol bound to gold, Au-S | 162.5 eV |
| Thiol, R-SH | ~164 eV |
| Na2(SO3)2 | 166.5 eV |
| Metal sulfate | ~169 eV |
Charge referenced to adventitious C1s peak at 284.8 eV.
N/A
Symbol: S
Date of discovery: ancient
Name origin: Sanskrit sulvari, arabic sufra
Appearance: lemon yellow
Discoverer: unknown
Obtained from: pure form or as sulfide/sulfate minerals
Melting point: 115.21 K
Boiling point: 444.6 K
Density[kg/m3]: (alpha)2.07
Molar volume: 15.53 × 10-6 m3/mol
Protons/Electrons: 16
Neutrons: 16
Shell structure: 2,8,6
Electron configuration: [Ne]3s23p4
Oxidation state: -1,±2,4,6
Crystal structure: orthorhombic
As a minor component of fats, body fluids, and skeletal materials, this pale yellow element is essential to human life. Sulfur is found in meteorites, volcanoes, and hot springs. Even one of Jupiter’s moons owes it’s colors to various forms of sulfur. An important manufactured chemical, sulfuric acid, is produced using sulfur. Sulfuric acid is commonly known as battery acid. In high concentrations, hydrogen sulfide can cause death by means of respiratory paralysis, and sulfur dioxide is known to be one of the causes of acid rain and a dangerous component in air pollution.
Products
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.