Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format
Thermo Scientific™

Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format

Thermo Scientific Pierce Sulfo-SMCC, No-Weigh Format is a water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at oppositeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
221221 g
A3926810 x 2 mg
2232250 mg
Catalog number 22122
Price (USD)
3,310.00
Each
Quantity:
1 g
Price (USD)
3,310.00
Each
Thermo Scientific Pierce Sulfo-SMCC, No-Weigh Format is a water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 angstroms).

Features of Sulfo-SMCC, No-Weigh Format:

• Reactive groups: sulfo-NHS ester and maleimide
• Reactive towards: amino and sulfhydryl groups
• Water-soluble (compare to SMCC), so crosslinking can be done in physiologic solutions
• High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
• Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.

Sulfosuccinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate (Sulfo-SMCC) is a non-cleavable and membrane impermeable crosslinker. It contains an amine-reactive N-hydroxysuccinimide (NHS ester) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC and SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to ∼10 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.

Applications:
• Enzyme labeling of antibodies - both enzyme and antibody specificity can be preserved
• Create specific bioconjugates via one- or two-step crosslinking reactions
• Create sulfhydryl-reactive, maleimide-activated carrier proteins for coupling haptens

Specifications:
Sulfo-SMCC is manufactured to the highest specifications to produce the most specific bioconjugates and maleimide-activated proteins, ensure the integrity of your data and to provide you with the highest degree of crosslinking consistency. Each lot of Sulfo-SMCC is tested to meet the following minimum specifications.

• Identity: IR scan shows only peaks characteristic of the structure and functional groups of Sulfo-SMCC crosslinker
• Purity: ≥ 90% by quantitative NMR (the highest standard for crosslinker purity)
• Solubility: ≥ 10 mg/mL in DI water

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Pierce™ Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
SM(PEG)4 (PEGylated SMCC crosslinker)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionSulfo-SMCC
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)436.37
PEGylatedNo
Product LinePierce
Quantity1 g
Reactive MoietyMaleimide, Sulfo-NHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length8.3 Å
Water SolubleYes
Chemical ReactivityAmine-Sulfhydryl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard, Single-use, Premium-grade
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at -20°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)?

Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the shelf life of Sulfo-SMCC (Cat. No. 22322)?

Sulfo-SMCC is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (desiccated at -20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the difference between SMCC and Sulfo-SMCC?

SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm (8.3 angstroms). Sulfo-SMCC is a similar compound but it has a sulfonate group on the NHS ring which makes it water-soluble whereas SMCC is water-insoluble and needs to be dissolved first in an organic solvent like DMF or DMSO. Subsequent dilution into aqueous reaction buffer is generally possible, and most protein reactants will remain soluble if the final concentration of organic solvent is less than 10%.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is the Thermo Scientific IP Lysis Buffer similar to RIPA buffer?

The Thermo Scientific IP Lysis Buffer (Cat. No. 87787) is a mammalian whole cell lysis buffer based on a modified RIPA buffer formulation without SDS. This moderate strength lysis buffer is able to effectively solubilize cellular proteins but does not liberate genomic DNA or disrupt protein complexes like RIPA buffer.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is the cyclohexane ring in Sulfo-SMCC crosslinker cis or trans?

The cyclohexane ring in Sulfo-SMCC crosslinker is trans.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (5)

Citations & References
Abstract
Solid phase DNA amplification: characterisation of primer attachment and amplification mechanisms.
Authors:Adessi C, Matton G, Ayala G, Turcatti G, Mermod JJ, Mayer P, Kawashima E
Journal:Nucleic Acids Res
PubMed ID:11024189
Different chemical methods used to attach oligonucleotides by their 5'-end on a glass surface were tested in the framework of solid phase PCR where surface-bound instead of freely-diffusing primers are used to amplify DNA. Each method was first evaluated for its capacity to provide a high surface coverage of oligonucleotides ... More
Substrate-induced conformational change in bacterial complex I.
Authors:Mamedova AA, Holt PJ, Carroll J, Sazanov LA
Journal:J Biol Chem
PubMed ID:15037611
'The mechanism coupling electron transfer and proton pumping in respiratory complex I (NADH-ubiquinone oxidoreductase) has not been established, but it has been suggested that it involves conformational changes. Here, the influence of substrates on the conformation of purified complex I from Escherichia coli was studied by cross-linking and electron microscopy. ... More
Chondrex: new marker of joint disease.
Authors:Harvey S, Weisman M, O'Dell J, Scott T, Krusemeier M, Visor J, Swindlehurst C
Journal:Clin Chem
PubMed ID:9510855
Chondrex, a major secretory protein of human chondrocytes and synovial fibroblasts, is increased in serum of patients with joint and cartilage disease. We have developed a sandwich-type ELISA for quantifying chondrex in serum. The interassay CVs were 2.8-3.7% and the average within-run and total CVs were 3.6% and 5.4%, respectively. ... More
Development of sensitive immunoassays to detect amylin and amylin-like peptides in unextracted plasma.
Authors:Percy AJ, Trainor DA, Rittenhouse J, Phelps J, Koda JE
Journal:Clin Chem
PubMed ID:8605675
Amylin is a 37-amino-acid polypeptide synthesized in and secreted from pancreatic beta cells along with insulin. Its biological actions include the slowing and reduction of postmeal increases in plasma glucose concentrations. Studies of the basic amylin biology in humans have been hampered by the lack of a rapid, sensitive assay ... More
Sulfhydryl-based tumor antigen-carrier protein conjugates stimulate superior antitumor immunity against B cell lymphomas.
Authors:Betting DJ, Kafi K, Abdollahi-Fard A, Hurvitz SA, Timmerman JM
Journal:J Immunol
PubMed ID:18768870
Therapeutic vaccination of B cell lymphoma patients with tumor-specific Ig (idiotype, or Id) chemically coupled to the immunogenic foreign carrier protein keyhole limpet hemocyanin (KLH) using glutaraldehyde has shown promising results in early clinical trials, and phase III trials are underway. However, glutaraldehyde Id-KLH vaccines fail to elicit anti-Id immune ... More