BMPS (N-β-maleimidopropyl-oxysuccinimide ester)
Thermo Scientific™
BMPS (N-β-maleimidopropyl-oxysuccinimide ester)
Thermo Scientific Pierce BMPS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of aRead more
Have Questions?
Catalog NumberQuantity
2229850 mg
Catalog number 22298
Price (USD)
175.00
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
175.00
Each
Thermo Scientific Pierce BMPS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short spacer arm (5.9 Å).
Features of BMPS:
• Reactive groups: NHS ester and maleimide
• Reactive towards: amino and sulfhydryl groups
• C3-homolog of aliphatic spacer series which includes AMAS, BMPS, GMBS and EMCS
• NHS ester end couples with primary amines at pH 7-9, forming stable amide bonds
• Maleimide reacts with -SH groups at pH 6.5-7.5, forming stable thioether linkages
• Non-cleavable
• Water-insoluble (dissolve first in DMF or DMSO)
• Short aliphatic spacer has low potential for eliciting an immune response
Product References:
Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionBMPS
FormSolid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)266.21
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietyMaleimide, NHS Ester
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length5.9 Å
Water SolubleNo
Chemical ReactivityAmine-Sulfhydryl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at 4°C.
Frequently asked questions (FAQs)
Can you provide the shelf-life for BMPS (N-β-maleimidopropyl-oxysuccinimide ester)?
BMPS (N-β-maleimidopropyl-oxysuccinimide ester) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Citations & References (1)
Citations & References
Abstract
Spatial and dynamic interactions between phospholamban and the canine cardiac Ca2+ pump revealed with use of heterobifunctional cross-linking agents.
Authors:Chen Z, Stokes DL, Rice WJ, Jones LR
Journal:J Biol Chem
PubMed ID:12972413
Heterobifunctional thiol to amine cross-linking agents were used to gain new insights on the dynamics and conformational factors governing the interaction between the cardiac Ca2+ pump (SERCA2a) and phospholamban (PLB). PLB is a small protein inhibitor of SERCA2a that reduces enzyme affinity for Ca2+ and thereby regulates cardiac contractility. We ... More