Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03302831_pri
          Assay Name:
          hsa-mir-148a
          Stem-loop ID:
          hsa-mir-148a
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.7: 25949919 - 25949986 [-] on Build GRCh38
          Stem-loop Location:
          Chr.7: 25949919 - 25949986 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-148a-3p hsa-miR-148a-5p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03302831_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          65

          Chromosome Location:

          Chr. 7 - 25950222 - 25950222 [-] on 25950222 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-148a-3p MIMAT0000243
          Human hsa-miR-148a-5p MIMAT0004549

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-148a
          Stem-loop Accession # MI0000253
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000243
          miRBase ID: hsa-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38

          miRBase Accession #: MIMAT0004549
          miRBase ID: hsa-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38

          Gene Family ID MIPF0000056, mir-148
          Distance to Amplicon 201 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10263
          Ambion® Pre-miR™ miRNA Precursor : PM10263
          TaqMan™ MicroRNA Assay : 000470
          mirVana® miRNA inhibitor : MH10263
          mirVana® miRNA mimic : MC10263
          TaqMan™ Advanced miRNA Assay : mmu477814_mir
          TaqMan™ Advanced miRNA Assay : 477814_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12683
          Ambion® Pre-miR™ miRNA Precursor : PM12683
          TaqMan™ MicroRNA Assay : 002134
          mirVana® miRNA inhibitor : MH12683
          mirVana® miRNA mimic : MC12683
          TaqMan™ Advanced miRNA Assay : 478718_mir
          TaqMan™ Advanced miRNA Assay : mmu481549_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch