Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10263
          Assay Name: hsa-miR-148a-3p
          miRBase Accession Number: MI0000253
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 25949919 - 25949986 [-] on Build GRCh38
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Species: Human, Mouse, Anolis carolinensis, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Saimiri boliviensis, Sheep, Taeniopygia guttata, Tupaia chinensis, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10263

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000056, mir-148

          Mature miRNA Sequence:

          UCAGUGCACUACAGAACUUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-148a-3p MIMAT0000243
          Mouse mmu-miR-148a-3p MIMAT0000516
          Anolis carolinensis aca-miR-148a-3p MIMAT0021781
          Bovine bta-miR-148a MIMAT0003522
          Callithrix jacchus cja-miR-148a MIMAT0039461
          Chicken gga-miR-148a-3p MIMAT0001120
          View More Rows
          Chrysemys picta cpi-miR-148a-3p MIMAT0037783
          Columba livia cli-miR-148a-3p MIMAT0038543
          Cricetulus griseus cgr-miR-148a MIMAT0041133
          Dasypus novemcinctus dno-miR-148a-3p MIMAT0047670
          Dog cfa-miR-148a MIMAT0006622
          Goat chi-miR-148a-3p MIMAT0035977
          Guinea pig cpo-miR-148a-3p MIMAT0047023
          Horse eca-miR-148a MIMAT0012935
          Monodelphis domestica mdo-miR-148-3p MIMAT0012737
          Ornithorhynchus anatinus oan-miR-148-3p MIMAT0007138
          Pan troglodytes ptr-miR-148a MIMAT0008041
          Pig ssc-miR-148a-3p MIMAT0002124
          Pongo pygmaeus ppy-miR-148a MIMAT0015771
          Pteropus alecto pal-miR-148a-3p MIMAT0039819
          Python bivittatus pbv-miR-148a-3p MIMAT0038965
          Rabbit ocu-miR-148a-3p MIMAT0048260
          Rhesus monkey mml-miR-148a-3p MIMAT0006208
          Saimiri boliviensis sbo-miR-148a MIMAT0049486
          Sheep oar-miR-148a MIMAT0030033
          Taeniopygia guttata tgu-miR-148a-3p MIMAT0014503
          Tupaia chinensis tch-miR-148a-3p MIMAT0036611
          Xenopus tropicalis xtr-miR-148a MIMAT0003664

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-148a
          Stem-loop Accession # MI0000253
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000243
          miRBase ID: hsa-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-148a
          Stem-loop Accession # MI0000550
          Stem-loop Sequence
          AGCCAGUUUGGUCUUUUGAGACAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAGAGGCUGUGGUC
          Chromosome Location Chr. 6 - 51269812 - 51269910 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000516
          miRBase ID: mmu-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: Chr. 6 - 51269812 - 51269910 [-] on Build GRCm38

          Anolis carolinensis

          Stem-loop ID aca-mir-148a
          Stem-loop Accession # MI0018759
          Stem-loop Sequence
          CAGAUUUGGUCUCUGGAAGCAAAGUUCUGUGACACUCGGACUCUGAUUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCGCCGGGGGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021781
          miRBase ID: aca-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-148a
          Stem-loop Accession # MI0004737
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003522
          miRBase ID: bta-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-148a
          Stem-loop Accession # MI0031985
          Stem-loop Sequence
          GGUCUUUUGAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAGAGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039461
          miRBase ID: cja-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-148a
          Stem-loop Accession # MI0001189
          Stem-loop Sequence
          GAAGCAAAGUUCUGUGACACUCAGACUCUGGUUACGAUAGCAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001120
          miRBase ID: gga-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-148a
          Stem-loop Accession # MI0029493
          Stem-loop Sequence
          GAUUUAGUCUCUUGAAGCAAAGUUCUGUGACACUCAGACUCUGAUUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUUUGGGGGCUGUAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037783
          miRBase ID: cpi-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-148a
          Stem-loop Accession # MI0029984
          Stem-loop Sequence
          GGAGCCUCCGGAAGCAAAGUUCUGUGACACUCCGACUCUGGGUACGAUAGCAGUCAGUGCACUACAGAACUUUGUCUCCGGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038543
          miRBase ID: cli-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-148a
          Stem-loop Accession # MI0033061
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUUCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041133
          miRBase ID: cgr-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-148a
          Stem-loop Accession # MI0038997
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047670
          miRBase ID: dno-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-148a
          Stem-loop Accession # MI0008018
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006622
          miRBase ID: cfa-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-148a
          Stem-loop Accession # MI0030636
          Stem-loop Sequence
          CAGUUUGGUCUUUUGAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035977
          miRBase ID: chi-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-148a
          Stem-loop Accession # MI0038656
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047023
          miRBase ID: cpo-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-148a
          Stem-loop Accession # MI0012689
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012935
          miRBase ID: eca-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-148
          Stem-loop Accession # MI0012493
          Stem-loop Sequence
          GAGGCAAAGUUCUGGGACACUCAGACUCUGAUUAGGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012737
          miRBase ID: mdo-miR-148-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-148
          Stem-loop Accession # MI0006868
          Stem-loop Sequence
          AGUCUUUUGAGGCAAAGUUCUGUGACACUCAGACUCUGAUUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCCAGAGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007138
          miRBase ID: oan-miR-148-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-148a
          Stem-loop Accession # MI0008547
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008041
          miRBase ID: ptr-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-148a
          Stem-loop Accession # MI0002418
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002124
          miRBase ID: ssc-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-148a
          Stem-loop Accession # MI0014837
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015771
          miRBase ID: ppy-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-148a
          Stem-loop Accession # MI0032345
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039819
          miRBase ID: pal-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-148a
          Stem-loop Accession # MI0030247
          Stem-loop Sequence
          AGUUUUGGUUUCUGGAAGCAAAGUUCUGUGACACUCGGACUCUGAUUAUAAUAGAAGUCAGUGCACUACAGAACUUUGUCUCCAGGGGCUGCAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038965
          miRBase ID: pbv-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-148a
          Stem-loop Accession # MI0039307
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUACGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048260
          miRBase ID: ocu-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-148a
          Stem-loop Accession # MI0007638
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006208
          miRBase ID: mml-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-148a
          Stem-loop Accession # MI0040246
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049486
          miRBase ID: sbo-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-148a
          Stem-loop Accession # MI0025254
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030033
          miRBase ID: oar-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-148a
          Stem-loop Accession # MI0013693
          Stem-loop Sequence
          CCCCGGCGCCGGUACCGGCGGAGGACUGGAGCCUCCGGAAGCAAAGUUCUGUGACACUCCGACUCUGGGUACGAUAGCAGUCAGUGCACUACAGAACUUUGUCUCCGGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014503
          miRBase ID: tgu-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-148a
          Stem-loop Accession # MI0031281
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036611
          miRBase ID: tch-miR-148a-3p
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-148a
          Stem-loop Accession # MI0004905
          Stem-loop Sequence
          UAGUCUUUUAAAUCAAAGUUCUGUGACACUUAGACUCUGAAUAUGAUAGCAGUCAGUGCACUACAGAACUUUGUUUUGGGAGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003664
          miRBase ID: xtr-miR-148a
          Mature miRNA Sequence: UCAGUGCACUACAGAACUUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302831_pri
          TaqMan™ Pri-miRNA Assay : Mm03306713_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10263
          Ambion® Pre-miR™ miRNA Precursor : PM10263
          TaqMan™ MicroRNA Assay : 000470
          mirVana® miRNA mimic : MC10263
          TaqMan™ Advanced miRNA Assay : mmu477814_mir
          TaqMan™ Advanced miRNA Assay : 477814_mir


          miRBase Alias:

          Human: hsa-miR-148a (v17)
          Mouse: mmu-miR-148a (v17)
          Aeromonas caviae Ae398: aca-miR-148a (v18)
          Gallus gallus: gga-miR-148a (v19)
          Macaca mulatta: mml-miR-148a (v19)
          Monodelphis domestica: mdo-miR-148 (v19)
          Ochrobactrum anthropi: oan-miR-148 (v18)
          Staphylococcus schleiferi: ssc-miR-148a (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-148 (v19)
          Trabulsiella guamensis ATCC 49490: tgu-miR-148-3p (v21)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch