Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002134
          Assay Name: hsa-miR-148a*
          miRBase Accession Number: MI0000253
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 25949919 - 25949986 [-] on Build GRCh38
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Species: Human, Mouse, Dasypus novemcinctus, Goat, Guinea pig, Pig, Rabbit, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002134
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000056, mir-148

          Mature miRNA Sequence:

          AAAGUUCUGAGACACUCCGACU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-148a-5p MIMAT0004549
          Mouse mmu-miR-148a-5p MIMAT0004617
          Dasypus novemcinctus dno-miR-148a-5p MIMAT0047669
          Goat chi-miR-148a-5p MIMAT0035976
          Guinea pig cpo-miR-148a-5p MIMAT0047022
          Pig ssc-miR-148a-5p MIMAT0022920
          View More Rows
          Rabbit ocu-miR-148a-5p MIMAT0048259
          Rhesus monkey mml-miR-148a-5p MIMAT0026825

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-148a
          Stem-loop Accession # MI0000253
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004549
          miRBase ID: hsa-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-148a
          Stem-loop Accession # MI0000550
          Stem-loop Sequence
          AGCCAGUUUGGUCUUUUGAGACAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAGAGGCUGUGGUC
          Chromosome Location Chr. 6 - 51269812 - 51269910 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004617
          miRBase ID: mmu-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: Chr. 6 - 51269812 - 51269910 [-] on Build GRCm38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-148a
          Stem-loop Accession # MI0038997
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047669
          miRBase ID: dno-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-148a
          Stem-loop Accession # MI0030636
          Stem-loop Sequence
          CAGUUUGGUCUUUUGAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035976
          miRBase ID: chi-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-148a
          Stem-loop Accession # MI0038656
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047022
          miRBase ID: cpo-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-148a
          Stem-loop Accession # MI0002418
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022920
          miRBase ID: ssc-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-148a
          Stem-loop Accession # MI0039307
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUACGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048259
          miRBase ID: ocu-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-148a
          Stem-loop Accession # MI0007638
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026825
          miRBase ID: mml-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool B v3.0
          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Cards:

          TaqMan® Rodent MicroRNA B Cards v3.0
          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panels:

          TaqMan® OpenArray® Human MicroRNA Panel
          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306713_pri
          TaqMan™ Pri-miRNA Assay : Hs03302831_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12683
          Ambion® Pre-miR™ miRNA Precursor : PM12683
          mirVana® miRNA inhibitor : MH12683
          mirVana® miRNA mimic : MC12683
          TaqMan™ Advanced miRNA Assay : 478718_mir
          TaqMan™ Advanced miRNA Assay : mmu481549_mir
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : TaqManTM Human MicroRNA Array B v3
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v2.0


          miRBase Alias:

          Human: hsa-miR-148a* (v17)
          Mouse: mmu-miR-148a* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch