Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12683
          Assay Name: hsa-miR-148a-5p
          miRBase Accession Number: MI0000253
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 25949919 - 25949986 [-] on Build GRCh38
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Species: Human, Mouse, Dasypus novemcinctus, Goat, Guinea pig, Pig, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12683

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000056, mir-148

          Mature miRNA Sequence:

          AAAGUUCUGAGACACUCCGACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-148a-5p MIMAT0004549
          Mouse mmu-miR-148a-5p MIMAT0004617
          Dasypus novemcinctus dno-miR-148a-5p MIMAT0047669
          Goat chi-miR-148a-5p MIMAT0035976
          Guinea pig cpo-miR-148a-5p MIMAT0047022
          Pig ssc-miR-148a-5p MIMAT0022920
          View More Rows
          Rabbit ocu-miR-148a-5p MIMAT0048259
          Rhesus monkey mml-miR-148a-5p MIMAT0026825

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-148a
          Stem-loop Accession # MI0000253
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004549
          miRBase ID: hsa-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: Chr. 7 - 25949919 - 25949986 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-148a
          Stem-loop Accession # MI0000550
          Stem-loop Sequence
          AGCCAGUUUGGUCUUUUGAGACAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAGAGGCUGUGGUC
          Chromosome Location Chr. 6 - 51269812 - 51269910 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004617
          miRBase ID: mmu-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: Chr. 6 - 51269812 - 51269910 [-] on Build GRCm38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-148a
          Stem-loop Accession # MI0038997
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047669
          miRBase ID: dno-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-148a
          Stem-loop Accession # MI0030636
          Stem-loop Sequence
          CAGUUUGGUCUUUUGAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035976
          miRBase ID: chi-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-148a
          Stem-loop Accession # MI0038656
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047022
          miRBase ID: cpo-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-148a
          Stem-loop Accession # MI0002418
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAAUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022920
          miRBase ID: ssc-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-148a
          Stem-loop Accession # MI0039307
          Stem-loop Sequence
          AAAGUUCUGAGACACUCCGACUCUGAGUACGAUAGAAGUCAGUGCACUACAGAACUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048259
          miRBase ID: ocu-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-148a
          Stem-loop Accession # MI0007638
          Stem-loop Sequence
          GAGGCAAAGUUCUGAGACACUCCGACUCUGAGUAUGAUAGAAGUCAGUGCACUACAGAACUUUGUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026825
          miRBase ID: mml-miR-148a-5p
          Mature miRNA Sequence: AAAGUUCUGAGACACUCCGACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306713_pri
          TaqMan™ Pri-miRNA Assay : Hs03302831_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12683
          Ambion® Pre-miR™ miRNA Precursor : PM12683
          TaqMan™ MicroRNA Assay : 002134
          mirVana® miRNA mimic : MC12683
          TaqMan™ Advanced miRNA Assay : 478718_mir
          TaqMan™ Advanced miRNA Assay : mmu481549_mir


          miRBase Alias:

          Human: hsa-miR-148a* (v17)
          Mouse: mmu-miR-148a* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0