Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12471
          Assay Name: hsa-miR-323b-5p
          miRBase Accession Number: MI0014206
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101056219 - 101056300 [+] on Build GRCh38
          Mature miRNA Sequence: AGGUUGUCCGUGGUGAGUUCGCA
          Species: Human, Bovine, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12471

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AGGUUGUCCGUGGUGAGUUCGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-323b-5p MIMAT0001630
          Bovine bta-miR-453 MIMAT0009325
          Pan troglodytes ptr-miR-453 MIMAT0008155
          Pongo pygmaeus ppy-miR-453 MIMAT0015898
          Rhesus monkey mml-miR-323b-5p MIMAT0028354

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-323b
          Stem-loop Accession # MI0014206
          Stem-loop Sequence
          UGGUACUCGGAGGGAGGUUGUCCGUGGUGAGUUCGCAUUAUUUAAUGAUGCCCAAUACACGGUCGACCUCUUUUCGGUAUCA
          Chromosome Location Chr. 14 - 101056219 - 101056300 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001630
          miRBase ID: hsa-miR-323b-5p
          Mature miRNA Sequence: AGGUUGUCCGUGGUGAGUUCGCA
          Chromosome Location: Chr. 14 - 101056219 - 101056300 [+] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-453
          Stem-loop Accession # MI0009839
          Stem-loop Sequence
          GCGGGAACGCCACCAGCAGCACUGCCUCACGGUACUUGGAGGGAGGUUGUCCGUGGUGAGUUCGCAUUAUUUAAUGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009325
          miRBase ID: bta-miR-453
          Mature miRNA Sequence: AGGUUGUCCGUGGUGAGUUCGCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-453
          Stem-loop Accession # MI0008677
          Stem-loop Sequence
          CAGGAAUGCUGCGAGCAGUGCCACCUCAUGGUACUCGGAGGGAGGUUGUCCGUGGUGAGUUCGCAUUAUUUAAUGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008155
          miRBase ID: ptr-miR-453
          Mature miRNA Sequence: AGGUUGUCCGUGGUGAGUUCGCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-453
          Stem-loop Accession # MI0014954
          Stem-loop Sequence
          GCAGGAAUGCUGCGAGCAGUGCCACCUCAUGGUACUCGGAGGGAGGUUGUCCGUGGUGAGUUCGCAUUAUUUAAUGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015898
          miRBase ID: ppy-miR-453
          Mature miRNA Sequence: AGGUUGUCCGUGGUGAGUUCGCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-323b
          Stem-loop Accession # MI0023688
          Stem-loop Sequence
          AGGUUGUCCGUGGUGAGUUCGCAUUAUUUAAUGAUGCCCAAUACACGGUCGACCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028354
          miRBase ID: mml-miR-323b-5p
          Mature miRNA Sequence: AGGUUGUCCGUGGUGAGUUCGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303761_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12471
          Ambion® Pre-miR™ miRNA Precursor : PM12471
          TaqMan™ MicroRNA Assay : 002318
          mirVana® miRNA inhibitor : MH12471
          TaqMan™ Advanced miRNA Assay : 478826_mir


          miRBase Alias:

          Macaca mulatta: mml-miR-453 (v21)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline