Search
Search

| Assay Name: | hsa-miR-323b-5p |
|---|---|
| miRBase Accession Number: | MI0014206 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUUGUCCGUGGUGAGUUCGCA |
| Species: | Human, Bovine, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUGUCCGUGGUGAGUUCGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-323b-5p | MIMAT0001630 |
| Bovine | bta-miR-453 | MIMAT0009325 |
| Pan troglodytes | ptr-miR-453 | MIMAT0008155 |
| Pongo pygmaeus | ppy-miR-453 | MIMAT0015898 |
| Rhesus monkey | mml-miR-323b-5p | MIMAT0028354 |