Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13109
          Assay Name: hsa-miR-122-3p
          miRBase Accession Number: MI0000442
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 58451074 - 58451158 [+] on Build GRCh38
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Species: Human, Anolis carolinensis, Chicken, Columba livia, Dasypus novemcinctus, Guinea pig, Python bivittatus, Rabbit, Salmo salar
          Product Type: mirVana® miRNA mimic
          Assay ID MC13109

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000095, mir-122

          Mature miRNA Sequence:

          AACGCCAUUAUCACACUAAAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-122-3p MIMAT0004590
          Anolis carolinensis aca-miR-122-3p MIMAT0021724
          Chicken gga-miR-122-3p MIMAT0026550
          Columba livia cli-miR-122-3p MIMAT0038492
          Dasypus novemcinctus dno-miR-122-3p MIMAT0047611
          Guinea pig cpo-miR-122-3p MIMAT0046962
          View More Rows
          Python bivittatus pbv-miR-122-3p MIMAT0038910
          Rabbit ocu-miR-122-3p MIMAT0048202
          Salmo salar ssa-miR-122-2-3p MIMAT0032303

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-122
          Stem-loop Accession # MI0000442
          Stem-loop Sequence
          CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC
          Chromosome Location Chr. 18 - 58451074 - 58451158 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004590
          miRBase ID: hsa-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: Chr. 18 - 58451074 - 58451158 [+] on Build GRCh38

          Anolis carolinensis

          Stem-loop ID aca-mir-122
          Stem-loop Accession # MI0018719
          Stem-loop Sequence
          UCCUGCUGGAGCUGUGGAGUGUGACAAUGGUGUUUGUAUCCAAUCCGUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021724
          miRBase ID: aca-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-122-1
          Stem-loop Accession # MI0001277
          Stem-loop Sequence
          CAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026550
          miRBase ID: gga-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Stem-loop ID gga-mir-122-2
          Stem-loop Accession # MI0001280
          Stem-loop Sequence
          CAGAGCUAUGGAGUGUGACAAUGGUGUUUGUGUCCAAUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026550
          miRBase ID: gga-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-122
          Stem-loop Accession # MI0029953
          Stem-loop Sequence
          CUGCCCGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCUGGCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGGUAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038492
          miRBase ID: cli-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-122
          Stem-loop Accession # MI0038963
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCACAAUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047611
          miRBase ID: dno-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-122
          Stem-loop Accession # MI0038621
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046962
          miRBase ID: cpo-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-122
          Stem-loop Accession # MI0030210
          Stem-loop Sequence
          UUCUGCUGGAGCUUUGGAGUGUGACAAUGGUGUUUGUAUCCAAUCUCUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038910
          miRBase ID: pbv-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-122
          Stem-loop Accession # MI0039275
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048202
          miRBase ID: ocu-miR-122-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-122-2
          Stem-loop Accession # MI0026451
          Stem-loop Sequence
          UGGAGUGUGACAAUGGUGUUUGUGUUCUCUGCAAUCAAACGCCAUUAUCACACUAAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032303
          miRBase ID: ssa-miR-122-2-3p
          Mature miRNA Sequence: AACGCCAUUAUCACACUAAAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303072_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13109
          Ambion® Pre-miR™ miRNA Precursor : PM13109
          TaqMan™ MicroRNA Assay : 002130
          mirVana® miRNA inhibitor : MH13109
          TaqMan™ Advanced miRNA Assay : 477874_mir


          miRBase Alias:

          Human: hsa-miR-122* (v17)
          Aeromonas caviae Ae398: aca-miR-122* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Thermo Fisher Scientific India Pvt Ltd.

          Registered office address:
          402, 403, 404, Delphi 'B' Wing,
          Hiranandani Business Park,
          Powai, Mumbai – 400 076

          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100MH2000PTC126872

          Invitrogen BioServices (India) Pvt Ltd

          Registered office address:
          First Technology Place, No 3, EPIP,
          Whitefield, Bangalore – 560 079

          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100KA2004PTC035330

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0