The affordable choice for fast, reliable nucleic acid and protein measurements
The affordable Thermo Scientific NanoDrop Lite Plus Microvolume Spectrophotometer enables any lab to assess purified DNA, RNA, and protein concentrations and purity ratios quickly and easily. While this entry-level instrument offers fewer features than our premium models, it delivers the same fast, accurate, reliable measurements from just 1–2 µL of sample using trusted NanoDrop technology. 
Compact, portable, and easy to use, the NanoDrop Lite Plus Spectrophotometer is well suited for quality check measurements in busy research labs and is a valuable teaching tool as well. 
NanoDrop Lite Plus Spectrophotometer features
The NanoDrop Lite Plus Spectrophotometer shares ultraviolet (UV) absorbance technology with our flagship NanoDrop Ultra Spectrophotometers and Fluorometers and NanoDrop Eight spectrophotometers, while offering a streamlined feature set for simplicity and affordability.
Calculates critical purity ratios
Measures purified DNA, RNA, and protein concentration up to 30 Abs and calculates A260/A280 and A260/A230 purity ratios.
Optional docking printer
Prints cryogenic labels.
Simple touchscreen interace
Portable instrument requires no attached PC.
The intuitive, easy-to-use NanoDrop Lite Plus touchscreen interface. (left) Home screen showing application categories; (right) sample measurement screen.
NanoDrop Lite Plus Spectrophotometer specifications
Attribute
Description
Absorbance accuracy
3% at 0.97 A, 302 nm, 23 ± 2C°
Absorbance expressed as Abs/mm measured at 25 °C
Absorbance range
0.04–30 Abs (10 mm equivalent)
Certifications/Compliance
UL/CSA and CE
Compatibility
Local Control Only
Concentration limits
dsDNA: 1,500 ng/μL; RNA: 1,200 ng/μL
BSA: 45 mg/mL; IgG: 21 mg/mL
Connections
USB 2.0 Port
Detection limits
dsDNA: 2.0 ng/μL; RNA: 1.6 ng/μL
BSA: 0.06 mg/mL; IgG: 0.03 mg/mL
Detector type
2048-element CMOS linear image sensor
Dimensions (W x D x H)
27 cm x 22 cm x 22 cm (10.6 in x 8.7 in x 8.7 in)
Display
LCD Touchscreen
For use with (Application)
Nucleic acid quantification, DNA quantification, RNA quantification, A280 protein quantification
Light Source
Xenon flashlamp
Measurement time
≤ 5 seconds
Pathlength
1.0 mm and 0.2 mm (automatically adjusting)
Power consumption
18 W (standby < 3 Watts)
Printer included
Cat. No. NDLPLUSGL: No
Cat. No. NDLPLUSPRGL: Yes
Sample volume
1–2 μL
Spectral resolution
≤ 1.8 nm (FWHM at Hg 254 nm)
Warranty
2 year
Wavelength accuracy
± 1 nm
Wavelength range
230 nm, 260 nm, 280 nm
Wavelength repeatability
Typical: 0.002 A (1.0 mm path) or 1%CV, whichever is greater
(SD of 20 individual measurements at 0.74 Abs)
Weight
Without printer 2.7 kg (6.0 lb); With printer: 3.2 kg (7.1 lb)
Voltage
100/240 V
NanoDrop Lite Plus Spectrophotometer ordering information
For information about accessories for the NanoDrop Lite Plus Spectrophotometer, see the Accessories tab.
For Research Use Only. Not for use in diagnostic procedures.