Search
As a cell scientist shaping the future of human health, ensuring the integrity of your work is paramount. Choose any Vios iDx CO₂ Incubator with confidence, knowing it is meticulously designed for optimal cell growth and minimal contamination risk.
The Vios iDx line of incubators combine our top CO2 incubator features. Watch the video and continue down the page to learn more.
Vios iDx 165 CO2 Incubator |
Vios iDx 165 CO2 Incubator with Cell Locker System |
|
| Individual chamber volume |
||
165 L |
165 L |
255 L |
| Sensors | ||
Choice of TC180 or IR180Si |
||
| Chamber material | ||
Choice of electropolished stainless steel or 100 % pure copper |
||
| Oxygen control | ||
Available as a factory-install option. Choice of 1 – 21 % control or 5 – 90 % control.
Vios iDx 165 and 255 CO2 Incubators with Tri-Gas 1 -21 % oxygen control (Americas only) |
||
| Stacking option | ||
Stack two 165 L incubators by combing the desired two incubators with: |
Stack two 255 L incubators by combining the desired two incubators with: |
|
Vios iDx CR 165 CO2 Incubator |
Vios iDx CR 255 CO2 Incubator |
|
| Specialized features | ||
Certified cleanroom compatible, ISO Class 5, GMP Grade A/B environments, FAT documentation package included |
Automated door for continuous hands free operation |
|
| Individual chamber volume | ||
165 L |
255 L |
255L |
| Sensors | ||
IR180SI |
IR180Si |
IR180Si |
| Chamber material | ||
Brushed stainless steel exterior casing with IP54 compliance |
Powder-coated cold-rolled steel or optional brushed stainless steel |
|
| Stacking option | ||
Stack two incubators by combining the desired two incubators with: |
Stack two incubators by combining the desired two incubators with: |
Speak to a specialist about your requirements.
|
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.