Search
Designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen.
These products have a 2 year warranty and are CE, cULus, and RoHS certified. Package contains power cord with adapters (when necessary) and a block lifter.
| 1 block Touch Screen Dry Bath | 2 block Touch Screen Dry Bath | 4 block Touch Screen Dry Bath | |
|---|---|---|---|
![]() | ![]() | ![]() | |
| Block capacity (sold separately) | 1 | 2 | 4 |
| Controller | PID digital | ||
| Temp range | Ambient +5° to 130°C (Ambient at 25°C) | ||
| Temp control accuracy | at 37°C ≤ ± 0.5°C | ||
| Temp uniformity | at 37°C ≤ ± 1°C | ||
| Timer range | 0-99:59 min or continuous | ||
| Dimensions | 14.7 x 7.9 x 3.9 in. (373 x 200 x 100mm) | 15.9 x 7.9 x 3.9 in. (403 x 200 x 100mm) | 21.1 x 7.9 x 3.9 in. (535 x 200 x 100mm) |
| Heat up time | ≤ 20 min 30°C to 130°C | ≤ 25 min 30°C to 130°C | |
| Weight | 7.5 lbs (3.4kg) | 8.2 lbs (3.7kg) | 11.2 lbs (5.1kg) |
| Voltage (50/60Hz) | 100-120V, 200-240V | 100-120V, 200-240V | 100-120V, 200-240V |
| Fuse Protector ø5×20 | 250V 2.5A | 250V 5A (120V) 250V 2.5A (240V) | 250V 8A (120V) 250V 3.15A (240V) |
| Order now | 88870007 (US) 88870010 (CN, EU, UK) | 88870008 (US) 88870011 (CN, EU, UK) | 88870009 (US) 88870012 (CN, EU, UK) |