Search
Expression materials and methods: The cDNA ORF of each gene was cloned into pDEST27 expression vector by Gateway cloning, which incorporates a GST tag to the N-terminal of each protein. The proteins were expressed by transfection of the cloned plasmids into Expi293F cell cultures in 96-deep well blocks according to the application note “Expi293 Expression System – Optimizing culture conditions to maximize protein yield and enable use of 96-well microtiter plates.” Expressed proteins were purified from cell lysates using 96-well glutathione-affinity plates and quantitated on a Caliper LabChip 90 (LC90) instrument. Limit of detection of the LC90 assay is 1 mg/L. Protein yields below this limit of detection are shown as <1. Data are presented in order from highest to lowest expression yield. Click column headings to sort data differently, or search to filter table.
No records were found matching your criteria
| Gene symbol | Description | Nucleotide sequence | Protein sequence | Expi293 system yield (mg/L) |
|---|---|---|---|---|
| PLEK | Pleckstrin, mRNA (cDNA clone MGC:17111 IMAGE:4341823), complete CDS | BC018549.1 | AAH18549.1 | 5610 |
| STAT3 | Signal transducer and activator of transcription 3 (acute-phase response factor) (STAT3), transcript variant 1, mRNA | NM_139276.2 | NP_644805.1 | 2139 |
| BID | BH3 interacting domain death agonist (BID), transcript variant 2, mRNA | NM_001196.3 | NP_001187.1 | 972 |
| RAB38 | RAB38, member RAS oncogene family (RAB38), mRNA | NM_022337.2 | NP_071732.1 | 702 |
| S100A4 | S100 calcium binding protein A4 (S100A4), transcript variant 1, mRNA | NM_002961.2 | NP_002952.1 | 609 |
| NQO1 | NAD(P)H dehydrogenase, quinone 1 (NQO1), transcript variant 1, mRNA | NM_000903.2 | NP_000894.1 | 525 |
| PSAP | Prosaposin (PSAP), transcript variant 1, mRNA | NM_002778.2 | NP_002769.1 | 524 |
| TFAP2A | Transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha) (TFAP2A), transcript variant 2, mRNA | NM_001032280.2 | NP_001027451.1 | 478 |
| NELFCD | TH1-like (Drosophila), mRNA (cDNA clone MGC:22971 IMAGE:4860894), complete CDS | BC014952.1 | AAH14952.1 | 411 |
| GRB2 | Growth factor receptor-bound protein 2 (GRB2), transcript variant 1, mRNA | NM_002086.4 | NP_002077.1 | 333 |
| ID1 | Inhibitor of DNA binding 1, dominant negative helix-loop-helix protein (ID1), transcript variant 1, mRNA | NM_002165.3 | NP_002156.2 | 295 |
| S100A6 | S100 calcium binding protein A6 (S100A6), mRNA | NM_014624.3 | NP_055439.1 | 246 |
| EGFR | Epidermal growth factor Receptor (EGFR), transcript variant 3, mRNA | NM_201283.1 | NP_958440.1 | 206 |
| ILK | Integrin-linked kinase (ILK), transcript variant 2, mRNA | NM_001014794.1 | NP_001014794.1 | 199 |
| S100P | S100 calcium binding protein P (S100P), mRNA | NM_005980.2 | NP_005971.1 | 197 |
| MTAP | Methylthioadenosine phosphorylase, mRNA (cDNA clone MGC:33067 IMAGE:4820938), complete CDS | BC026106.1 | AAH26106.1 | 182 |
| OAT | Ornithine aminotransferase (OAT), nuclear gene encoding mitochondrial protein, transcript variant 1, mRNA | NM_000274.3 | NP_000265.1 | 179 |
| C12orf57 | Chromosome 12 open reading frame 57 (C12orf57), mRNA | NM_138425.2 | NP_612434.1 | 175 |
| NPRL2 | Tumor suppressor candidate 4, mRNA (cDNA clone MGC:22898 IMAGE:4068981), complete CDS | BC021984.1 | AAH21984.1 | 175 |
| NDRG1 | N-myc downstream regulated 1 (NDRG1), transcript variant 2, mRNA | NM_006096.3 | NP_006087.2 | 162 |
| MSH2 | MutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli) (MSH2), transcript variant 1, mRNA | NM_000251.2 | NP_000242.1 | 159 |
| CALR | Calreticulin (CALR), mRNA | NM_004343.3 | NP_004334.1 | 148 |
| MVP | Major vault protein (MVP), transcript variant 2, mRNA | NM_005115.4 | NP_005106.2 | 139 |
| MAP2K4 | Mitogen-activated protein kinase kinase 4, mRNA (cDNA clone MGC:33126 IMAGE:5272439), complete CDS | BC036032.1 | AAH36032.1 | 138 |
| XRCC3 | X-ray repair complementing defective repair in Chinese hamster cells 3, mRNA (cDNA clone MGC:19630 IMAGE:4138588), complete CDS | BC011725.2 | AAH11725.1 | 138 |
| BUB1B | Budding uninhibited by benzimidazoles 1 homolog beta (yeast), mRNA (cDNA clone MGC:31884 IMAGE:4649881), complete CDS | BC018739.1 | AAH18739.1 | 135 |
| GNAS | GNAS complex locus (GNAS), transcript variant 3, mRNA | NM_080426.2 | NP_536351.1 | 125 |
| CD4 | CD4 molecule (CD4), transcript variant 1, mRNA | NM_000616.4 | NP_000607.1 | 108 |
| TP53 | Tumor protein p53, mRNA (cDNA clone MGC:646 IMAGE:3544714), complete CDS | BC003596.1 | AAH03596.1 | 100 |
| HNF1A | HNF1 homeobox A (HNF1A), mRNA | NM_000545.5 | NP_000536.5 | 84 |
| SULT1E1 | Sulfotransferase family 1E, estrogen-preferring, member 1, mRNA (cDNA clone MGC:34459 IMAGE:5210178), complete CDS | BC027956.1 | AAH27956.1 | 79 |
| CHI3L1 | Chitinase 3-like 1 (cartilage glycoprotein-39), mRNA (cDNA clone MGC:17199 IMAGE:4212419), complete CDS | BC038354.1 | AAH38354.1 | 76 |
| CSK | c-Src tyrosine kinase (CSK), transcript variant 1, mRNA | NM_004383.2 | NP_004374.1 | 72 |
| THBD | Thrombomodulin, mRNA (cDNA clone MGC:45302 IMAGE:5176531), complete CDS | BC035602.1 | AAH35602.2 | 71 |
| JAK2 | Janus kinase 2 (JAK2), mRNA | NM_004972.3 | NP_004963.1 | 69 |
| SEMA3B | Sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B, mRNA (cDNA clone MGC:18122 IMAGE:4153377), complete CDS | BC009113.1 | AAH09113.1 | 67 |
| HORMAD1 | HORMA domain containing 1 (HORMAD1), transcript variant 1, mRNA | NM_032132.4 | NP_115508.2 | 66 |
| TMEM37 | transmembrane protein 37, mRNA (cDNA clone MGC:50757 IMAGE:5221396), complete CDS | BC046362.1 | AAH46362.1 | 62 |
| ACTL8 | Actin-like 8, mRNA (cDNA clone MGC:24924 IMAGE:4933032), complete cds | BC028909.1 | AAH28909.1 | 56 |
| ICAM1 | Intercellular adhesion molecule 1 (ICAM1), mRNA | NM_000201.2 | NP_000192.2 | 54 |
| DHFR | Dihydrofolate reductase, mRNA (cDNA clone MGC:88273 IMAGE:6714220), complete CDS | BC070280.1 | AAH70280.1 | 47 |
| FEV | FEV (ETS oncogene family) (FEV), mRNA | NM_017521.2 | NP_059991.1 | 47 |
| C8orf4 | Chromosome 8 open reading frame 4 (C8orf4), mRNA | NM_020130.4 | NP_064515.1 | 46 |
| FABP1 | Fatty acid binding protein 1, liver (FABP1), mRNA | NM_001443.2 | NP_001434.1 | 43 |
| MMP10 | Matrix metallopeptidase 10 (stromelysin 2) (MMP10), mRNA | NM_002425.2 | NP_002416.1 | 41 |
| PTPN11 | Protein tyrosine phosphatase, non-receptor type 11 (PTPN11), transcript variant 2, mRNA | NM_080601.1 | NP_542168.1 | 40 |
| FBXW7 | F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase (FBXW7), transcript variant 2, mRNA | NM_018315.4 | NP_060785.2 | 39 |
| OR51E2 | Olfactory receptor, family 51, subfamily E, member 2 (OR51E2), mRNA | NM_030774.3 | NP_110401.1 | 39 |
| SMAD4 | SMAD family member 4 (SMAD4), mRNA | NM_005359.5 | NP_005350.1 | 35 |
| EML4 | Echinoderm microtubule associated protein like 4, mRNA (cDNA clone IMAGE:3868340), complete CDS | BC008685 | AAH08685 | 33 |
| ESR2 | Estrogen receptor 2 (ER beta), mRNA (cDNA clone MGC:26496 IMAGE:4825937), complete CDS | BC024181.2 | AAH24181.1 | 32 |
| MYCN | v-Myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) (MYCN), mRNA | NM_005378.4 | NP_005369.2 | 32 |
| SSX2 | Synovial sarcoma, X breakpoint 2 (SSX2), transcript variant 2, mRNA | NM_175698.1 | NP_783629.1 | 32 |
| CASC1 | Cancer susceptibility candidate 1 (CASC1), transcript variant 3, mRNA | NM_001082972.1 | NP_001076441.1 | 31 |
| GSTM1 | Glutathione S-transferase mu 1 (GSTM1), transcript variant 2, mRNA | NM_146421.2 | NP_666533.1 | 30 |
| TXN | Thioredoxin (TXN), transcript variant 1, mRNA | NM_003329.3 | NP_003320.2 | 30 |
| CYP2A6 | Cytochrome P450, family 2, subfamily A, polypeptide 6, mRNA (cDNA clone MGC:116920 IMAGE:40006064), complete CDS | BC096255.1 | AAH96255.1 | 29 |
| FANCC | Fanconi anemia, complementation group C (FANCC), transcript variant 1, mRNA | NM_000136.2 | NP_000127.2 | 29 |
| CDK4 | Cyclin-dependent kinase 4 (CDK4), mRNA | NM_000075.3 | NP_000066.1 | 28 |
| PIN1 | Peptidylprolyl cis/trans isomerase, NIMA-interacting 1 (PIN1), transcript variant 1, mRNA | NM_006221.3 | NP_006212.1 | 28 |
| PLAC1 | Placenta-specific 1 (PLAC1), mRNA | NM_021796.3 | NP_068568.1 | 28 |
| TNFRSF10B | Tumor necrosis factor receptor superfamily, member 10b, mRNA (cDNA clone MGC:5144 IMAGE:3458466), complete CDS | BC001281.1 | AAH01281.1 | 28 |
| CDCP1 | CUB domain containing protein 1 (CDCP1), transcript variant 2, mRNA | NM_178181.1 | NP_835488.1 | 27 |
| DLD | Dihydrolipoamide dehydrogenase (DLD), nuclear gene encoding mitochondrial protein, mRNA | NM_000108.3 | NP_000099.2 | 27 |
| XAGE1A | X antigen family, member 1A (XAGE1A), transcript variant a, mRNA | NM_001097592.2 | NP_001091061.2 | 27 |
| STAT5A | Signal transducer and activator of transcription 5A (STAT5A), mRNA | NM_003152.3 | NP_003143.2 | 26 |
| NRAS | Neuroblastoma RAS viral (v-ras) oncogene homolog (NRAS), mRNA | NM_002524.4 | NP_002515.1 | 25 |
| RHOC | Ras homolog family member C (RHOC), transcript variant 1, mRNA | NM_175744.4 | NP_786886.1 | 25 |
| APOD | Apolipoprotein D (APOD), mRNA | NM_001647.3 | NP_001638.1 | 24 |
| AFP | Alpha-fetoprotein (AFP), mRNA | NM_001134.1 | NP_001125.1 | 23 |
| BAD | BCL2-associated agonist of cell death (BAD), transcript variant 1, mRNA | NM_004322.3 | NP_004313.1 | 22 |
| KLK4 | Kallikrein-related peptidase 4, mRNA (cDNA clone MGC:96926 IMAGE:7262135), complete CDS | BC069429.1 | AAH69429.1 | 22 |
| RELB | V-rel reticuloendotheliosis viral oncogene homolog B (RELB), mRNA | NM_006509.3 | NP_006500.2 | 19 |
| TMPRSS2 | Transmembrane protease, serine 2 (TMPRSS2), transcript variant 2, mRNA | NM_005656.3 | NP_005647.3 | 19 |
| CXorf48 | Chromosome X open reading frame 48 (CXorf48), transcript variant 2, mRNA | NM_017863.2 | NP_060333.1 | 18 |
| DIABLO | Diablo, IAP-binding mitochondrial protein (DIABLO), nuclear gene encoding mitochondrial protein, transcript variant 1, mRNA | NM_019887.4 | NP_063940.1 | 18 |
| PDLIM5 | PDZ and LIM domain 5 (PDLIM5), transcript variant 5, mRNA | NM_001011516.2 | NP_001011516.1 | 18 |
| AGTR2 | Angiotensin II receptor, type 2 (AGTR2), mRNA | NM_000686.4 | NP_000677.2 | 17 |
| CYR61 | Cysteine-rich, angiogenic inducer, 61 (CYR61), mRNA | NM_001554.4 | NP_001545.2 | 17 |
| HLA-E | Major histocompatibility complex, class I, E, mRNA (cDNA clone MGC:1408 IMAGE:3140835), complete CDS | BC002578.2 | AAH02578.1 | 17 |
| HMGA1 | High mobility group AT-hook 1 (HMGA1), transcript variant 2, mRNA | NM_002131.3 | NP_002122.1 | 17 |
| PTH | Parathyroid hormone (PTH), mRNA | NM_000315.2 | NP_000306.1 | 17 |
| RET | Ret proto-oncogene, mRNA (cDNA clone IMAGE:3507569), complete CDS | BC003072 | AAH03072 | 17 |
| IGFBP2 | Insulin-like growth factor binding protein 2, 36 kDa (IGFBP2), mRNA | NM_000597.2 | NP_000588.2 | 16 |
| JAK3 | Janus kinase 3 (JAK3), mRNA | NM_000215.3 | NP_000206.2 | 16 |
| KLK3 | Kallikrein-related peptidase 3 (KLK3), transcript variant 1, mRNA | NM_001648.2 | NP_001639.1 | 16 |
| TGFB1 | Transforming growth factor, beta 1, mRNA (cDNA clone MGC:2323 IMAGE:3356605), complete CDS | BC001180.1 | AAH01180.1 | 16 |
| BCL2L2 | BCL2-like 2 (BCL2L2), transcript variant 1, mRNA | NM_004050.4 | NP_004041.1 | 15 |
| CYP17A1 | Cytochrome P450, family 17, subfamily A, polypeptide 1 (CYP17A1), mRNA | NM_000102.3 | NP_000093.1 | 15 |
| KRT5 | Keratin 5, mRNA (cDNA clone MGC:39301 IMAGE:5428482), complete CDS | BC024292.1 | AAH24292.1 | 15 |
| SMO | Smoothened, frizzled family receptor (SMO), mRNA | NM_005631.4 | NP_005622.1 | 15 |
| TYK2 | tyrosine kinase 2, mRNA (cDNA clone MGC:20776 IMAGE:4591726), complete CDS | BC014243.2 | AAH14243.1 | 15 |
| MT3 | Metallothionein 3 (MT3), mRNA | NM_005954.2 | NP_005945.1 | 14 |
| NCL | Nucleolin (NCL), mRNA | NM_005381.2 | NP_005372.2 | 14 |
| AMD1 | Adenosylmethionine decarboxylase 1 (AMD1), transcript variant 1, mRNA | NM_001634.4 | NP_001625.2 | 13 |
| KRT19 | Keratin 19, mRNA (cDNA clone MGC:15788 IMAGE:3504206), complete CDS | BC007628.1 | AAH07628.1 | 13 |
| EPHX1 | Epoxide hydrolase 1, microsomal (xenobiotic) (EPHX1), transcript variant 1, mRNA | NM_000120.3 | NP_000111.1 | 12 |
| GAGE12I | G antigen 12I (GAGE12I), mRNA | NM_001477.1 | NP_001468.1 | 12 |
| HLA-A | Major histocompatibility complex, class I, A, mRNA (cDNA clone MGC:17191 IMAGE:4157200), complete CDS | BC008611.1 | AAH08611.1 | 12 |
| LAMP1 | Lysosomal-associated membrane protein 1 (LAMP1), mRNA | NM_005561.3 | NP_005552.3 | 12 |
| LBR | Lamin B receptor (LBR), transcript variant 1, mRNA | NM_002296.3 | NP_002287.2 | 12 |
| PRLR | Prolactin receptor (PRLR), transcript variant 1, mRNA | NM_000949.5 | NP_000940.1 | 12 |
| ESR1 | Estrogen receptor 1 (ESR1), transcript variant 1, mRNA | NM_000125.3 | NP_000116.2 | 11 |
| ELAVL2 | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B) (ELAVL2), transcript variant 2, mRNA | NM_001171195.1 | NP_001164666.1 | 10 |
| RNF14 | Ring finger protein 14 (RNF14), transcript variant 2, mRNA | NM_183398.2 | NP_899645.1 | 10 |
| SS18 | Synovial sarcoma translocation, chromosome 18, mRNA (cDNA clone MGC:116875 IMAGE:40004909), complete CDS | BC096222.1 | AAH96222.1 | 10 |
| TWIST1 | Twist basic helix-loop-helix transcription factor 1 (TWIST1), mRNA | NM_000474.3 | NP_000465.1 | 10 |
| CEACAM1 | Carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein) (CEACAM1), transcript variant 6, mRNA | NM_001205344.1 | NP_001192273.1 | 9 |
| CT45A1 | Cancer/testis antigen family 45, member A1 (CT45A1), mRNA | NM_001017417.1 | NP_001017417.1 | 9 |
| FASN | Fatty acid synthase, mRNA (cDNA clone IMAGE:4299348), complete CDS | BC007909 | AAH07909 | 9 |
| MALT1 | Mucosa associated lymphoid tissue lymphoma translocation gene 1 (MALT1), transcript variant 2, mRNA | NM_173844.1 | NP_776216.1 | 9 |
| SFTPC | Surfactant protein C, mRNA (cDNA clone MGC:14509 IMAGE:4043169), complete CDS | BC005913.1 | AAH05913.1 | 9 |
| BCL2L1 | BCL2-like 1 (BCL2L1), nuclear gene encoding mitochondrial protein, transcript variant 1, mRNA | NM_138578.1 | NP_612815.1 | 8 |
| CAV1 | Caveolin 1, caveolae protein, 22kDa (CAV1), transcript variant 1, mRNA | NM_001753.4 | NP_001744.2 | 8 |
| FASLG | Fas ligand (TNF superfamily, member 6) (FASLG), mRNA | NM_000639.1 | NP_000630.1 | 8 |
| GDF15 | Growth differentiation factor 15 (GDF15), mRNA | NM_004864.2 | NP_004855.2 | 8 |
| OLR1 | Oxidized low density lipoprotein (lectin-like) receptor 1 (OLR1), transcript variant 1, mRNA | NM_002543.3 | NP_002534.1 | 8 |
| PCSK7 | Proprotein convertase subtilisin/kexin type 7, mRNA (cDNA clone MGC:12699 IMAGE:4125594), complete CDS | BC006357.2 | AAH06357.1 | 8 |
| EIF4EBP1 | Eukaryotic translation initiation factor 4E binding protein 1 (EIF4EBP1), mRNA | NM_004095.3 | NP_004086.1 | 7 |
| FYN | FYN oncogene related to SRC, FGR, YES (FYN), transcript variant 1, mRNA | NM_002037.5 | NP_002028.1 | 7 |
| IL10 | Interleukin 10 (IL10), mRNA | NM_000572.2 | NP_000563.1 | 7 |
| AMACR | Alpha-methylacyl-CoA racemase, mRNA (cDNA clone MGC:3743 IMAGE:2958112), complete CDS | BC009471.1 | AAH09471.1 | 6 |
| NHLH1 | Nescient helix loop helix 1 (NHLH1), mRNA | NM_005598.3 | NP_005589.1 | 6 |
| RAC1 | Ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) (RAC1), transcript variant Rac1, mRNA | NM_006908.4 | NP_008839.2 | 6 |
| RARB | Retinoic acid receptor, beta (RARB), transcript variant 1, mRNA | NM_000965.3 | NP_000956.2 | 6 |
| SFTPD | Surfactant protein D, mRNA (cDNA clone MGC:22626 IMAGE:4696173), complete CDS | BC022318.1 | AAH22318.1 | 6 |
| SNRPE | Small nuclear ribonucleoprotein polypeptide E (SNRPE), mRNA | NM_003094.2 | NP_003085.1 | 6 |
| CHRNA3 | Cholinergic receptor, nicotinic, alpha 3 (neuronal) (CHRNA3), transcript variant 1, mRNA | NM_000743.4 | NP_000734.2 | 5 |
| FEZF2 | FEZ family zinc finger 2, mRNA (cDNA clone MGC:26179 IMAGE:4794121), complete CDS | BC022464.1 | AAH22464.1 | 5 |
| IL6 | Interleukin 6 (interferon, beta 2) (IL6), mRNA | NM_000600.3 | NP_000591.1 | 5 |
| JUN | Jun proto-oncogene (JUN), mRNA | NM_002228.3 | NP_002219.1 | 5 |
| KLK10 | Kallikrein-related peptidase 10, mRNA (cDNA clone MGC:3667 IMAGE:3632557), complete CDS | BC002710.2 | AAH02710.1 | 5 |
| LEF1 | Lymphoid enhancer-binding factor 1 (LEF1), transcript variant 1, mRNA | NM_016269.4 | NP_057353.1 | 5 |
| ABCC5 | ATP-binding cassette, sub-family C (CFTR/MRP), member 5 (ABCC5), transcript variant 2, mRNA | NM_001023587.1 | NP_001018881.1 | 4 |
| CDC25B | Cell division cycle 25B (CDC25B), transcript variant 1, mRNA | NM_021873.2 | NP_068659.1 | 4 |
| CPM | Carboxypeptidase M (CPM), transcript variant 3, mRNA | NM_001005502.2 | NP_001005502.1 | 4 |
| CYP3A5 | Cytochrome P450, family 3, subfamily A, polypeptide 5 (CYP3A5), transcript variant 1, mRNA | NM_000777.3 | NP_000768.1 | 4 |
| DHCR24 | 24-dehydrocholesterol reductase (DHCR24), mRNA | NM_014762.3 | NP_055577.1 | 4 |
| ELF3 | E74-like factor 3 (ETS domain transcription factor, epithelial-specific ) (ELF3), transcript variant 2, mRNA | NM_001114309.1 | NP_001107781.1 | 4 |
| EWSR1 | Ewing sarcoma breakpoint region 1 (EWSR1), transcript variant 2, mRNA | NM_005243.3 | NP_005234.1 | 4 |
| KRT18 | Keratin 18 (KRT18), transcript variant 1, mRNA | NM_000224.2 | NP_000215.1 | 4 |
| SERPINA5 | Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5, mRNA (cDNA clone MGC:16086 IMAGE:3618167), complete CDS | BC008915.2 | AAH08915.1 | 4 |
| TFF1 | Trefoil factor 1 (TFF1), mRNA | NM_003225.2 | NP_003216.1 | 4 |
| TGFBR2 | Transforming growth factor, beta receptor II (70/80kDa) (TGFBR2), transcript variant 2, mRNA | NM_003242.5 | NP_003233.4 | 4 |
| FOLH1 | Folate hydrolase (prostate-specific membrane antigen) 1 (FOLH1), transcript variant 2, mRNA | NM_001014986.1 | NP_001014986.1 | 3 |
| FRZB | Frizzled-related protein (FRZB), mRNA | NM_001463.3 | NP_001454.2 | 3 |
| MAGEC2 | Melanoma antigen family C, 2, mRNA (cDNA clone MGC:13377 IMAGE:4133159), complete CDS | BC013318.1 | AAH13318.1 | 3 |
| PTN | Pleiotrophin (PTN), mRNA | NM_002825.5 | NP_002816.1 | 3 |
| RPS6 | Ribosomal protein S6 (RPS6), mRNA | NM_001010.2 | NP_001001.2 | 3 |
| SIAH1 | Siah E3 ubiquitin protein ligase 1 (SIAH1), transcript variant 2, mRNA | NM_001006610.1 | NP_001006611.1 | 3 |
| SSX1 | Synovial sarcoma, X breakpoint 1 (SSX1), mRNA | NM_005635.2 | NP_005626.1 | 3 |
| BAK1 | BCL2-antagonist/killer 1 (BAK1), mRNA | NM_001188.3 | NP_001179.1 | 2 |
| CCK | Cholecystokinin (CCK), transcript variant 1, mRNA | NM_000729.4 | NP_000720.1 | 2 |
| CLPTM1L | CLPTM1-like (CLPTM1L), mRNA | NM_030782.3 | NP_110409.2 | 2 |
| DES | Desmin (DES), mRNA | NM_001927.3 | NP_001918.3 | 2 |
| ELAVL4 | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D), mRNA (cDNA clone MGC:33705 IMAGE:5286347), complete CDS | BC036071.1 | AAH36071.1 | 2 |
| HOXB13 | Homeobox B13 (HOXB13), mRNA | NM_006361.5 | NP_006352.2 | 2 |
| NRG1 | Neuregulin 1, mRNA (cDNA clone IMAGE:3161700), complete CDS | BC007675 | AAH07675 | 2 |
| VIM | Vimentin (VIM), mRNA | NM_003380.3 | NP_003371.2 | 2 |
| WNT5A | Wingless-type MMTV integration site family, member 5A (WNT5A), transcript variant 1, mRNA | NM_003392.4 | NP_003383.2 | 2 |
| XPA | Xeroderma pigmentosum, complementation group A (XPA), transcript variant 1, mRNA | NM_000380.3 | NP_000371.1 | 2 |
| DLST | Dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex) (DLST), nuclear gene encoding mitochondrial protein, transcript variant 1, mRNA | NM_001933.4 | NP_001924.2 | 1 |
| IGFBP1 | Insulin-like growth factor binding protein 1, mRNA (cDNA clone MGC:30041 IMAGE:4800940), complete CDS | BC035263.1 | AAH35263.2 | 1 |
| IGFBP3 | Insulin-like growth factor binding protein 3, mRNA (cDNA clone MGC:2305 IMAGE:3506666), complete CDS | BC000013.2 | AAH00013.1 | 1 |
| IL8 | Interleukin 8 (IL8), mRNA | NM_000584.3 | NP_000575.1 | 1 |
| LCK | Lymphocyte-specific protein Tyrosine kinase, mRNA (cDNA clone MGC:17196 IMAGE:4341278), complete CDS | BC013200.1 | AAH13200.1 | 1 |
| MIA | Melanoma inhibitory activity (MIA), transcript variant 1, mRNA | NM_006533.3 | NP_006524.1 | 1 |
| MMP3 | Matrix metallopeptidase 3 (stromelysin 1, progelatinase) (MMP3), mRNA | NM_002422.3 | NP_002413.1 | 1 |
| MMP9 | matrix metallopeptidase 9 (gelatinase B, 92 kDa gelatinase, 92 kDa type IV collagenase), mRNA (cDNA clone MGC:12688 IMAGE:4054882), complete CDS | BC006093.1 | AAH06093.1 | 1 |
| PASD1 | PAS domain containing 1 (PASD1), mRNA | NM_173493.2 | NP_775764.2 | 1 |
| PRL | Prolactin (PRL), transcript variant 1, mRNA | NM_000948.5 | NP_000939.1 | 1 |
| TIMP3 | TIMP metallopeptidase inhibitor 3 (TIMP3), mRNA | NM_000362.4 | NP_000353.1 | 1 |
| ABCC4 | ATP-binding cassette, sub-family C (CFTR/MRP), member 4 (ABCC4), transcript variant 2, mRNA | NM_001105515.1 | NP_001098985.1 | <1 |
| BAX | BCL2-associated X protein (BAX), transcript variant alpha, mRNA | NM_138761.3 | NP_620116.1 | <1 |
| BCL2 | B-cell CLL/lymphoma 2 (BCL2), nuclear gene encoding mitochondrial protein, transcript variant alpha, mRNA | NM_000633.2 | NP_000624.2 | <1 |
| BIRC2 | Baculoviral IAP repeat containing 2 (BIRC2), transcript variant 1, mRNA | NM_001166.4 | NP_001157.1 | <1 |
| BMPR2 | Bone morphogenetic protein receptor, type II (serine/threonine kinase) (BMPR2), mRNA | NM_001204.6 | NP_001195.2 | <1 |
| BRD2 | Bromodomain containing 2 (BRD2), transcript variant 1, mRNA | NM_005104.3 | NP_005095.1 | <1 |
| C2 | Complement component 2, mRNA (cDNA clone IMAGE:5183067), complete CDS | BC029781 | AAH29781 | <1 |
| CALCR | Calcitonin receptor (CALCR), transcript variant 2, mRNA | NM_001742.3 | NP_001733.1 | <1 |
| CASP8 | Caspase 8, apoptosis-related cysteine peptidase (CASP8), transcript variant B, mRNA | NM_033355.3 | NP_203519.1 | <1 |
| CD200 | CD200 molecule, mRNA (cDNA clone MGC:33959 IMAGE:5301702), complete CDS | BC031103.1 | AAH31103.1 | <1 |
| CD63 | CD63 molecule (CD63), transcript variant 1, mRNA | NM_001780.5 | NP_001771.1 | <1 |
| CD82 | CD82 molecule, mRNA (cDNA clone MGC:1713 IMAGE:2959683), complete CDS | BC000726.2 | AAH00726.1 | <1 |
| CD99 | CD99 molecule (CD99), transcript variant 1, mRNA | NM_002414.3 | NP_002405.1 | <1 |
| CFLAR | CASP8 and FADD-like apoptosis regulator (CFLAR), transcript variant 1, mRNA | NM_003879.5 | NP_003870.4 | <1 |
| CGA | Glycoprotein hormones, alpha polypeptide, mRNA (cDNA clone MGC:13639 IMAGE:4043527), complete CDS | BC010957.1 | AAH10957.1 | <1 |
| CREB1 | cAMP responsive element Binding protein 1 (CREB1), transcript variant A, mRNA | NM_004379.3 | NP_004370.1 | <1 |
| CTCF | CCCTC-binding factor (zinc finger protein) (CTCF), transcript variant 1, mRNA | NM_006565.3 | NP_006556.1 | <1 |
| CXCL9 | Chemokine (C-X-C motif) ligand 9 (CXCL9), mRNA | NM_002416.1 | NP_002407.1 | <1 |
| CXorf61 | Chromosome X open reading frame 61 (CXorf61), mRNA | NM_001017978.3 | NP_001017978.1 | <1 |
| CYP3A4 | Cytochrome P450, family 3, subfamily A, polypeptide 4 (CYP3A4), transcript variant 1, mRNA | NM_017460.5 | NP_059488.2 | <1 |
| DDIT3 | DNA-damage-inducible transcript 3 (DDIT3), transcript variant 5, mRNA | NM_004083.5 | NP_004074.2 | <1 |
| DKK1 | Dickkopf 1 homolog (Xenopus laevis) (DKK1), mRNA | NM_012242.2 | NP_036374.1 | <1 |
| DLX4 | Distal-less homeobox 4 (DLX4), transcript variant 2, mRNA | NM_001934.3 | NP_001925.2 | <1 |
| EPHA2 | EPH receptor A2 (EPHA2), mRNA | NM_004431.3 | NP_004422.2 | <1 |
| FADD | Fas (TNFRSF6)-associated via death domain (FADD), mRNA | NM_003824.3 | NP_003815.1 | <1 |
| FANCB | Fanconi anemia, complementation group B (FANCB), transcript variant 1, mRNA | NM_001018113.1 | NP_001018123.1 | <1 |
| FAS | Fas (TNF receptor superfamily, member 6) (FAS), transcript variant 1, mRNA | NM_000043.4 | NP_000034.1 | <1 |
| FGFR1 | Fibroblast growth factor receptor 1, mRNA (cDNA clone MGC:9218 IMAGE:3896359), complete CDS | BC018128.1 | AAH18128.1 | <1 |
| FLT1 | Fms-related tyrosine kinase 1 (FLT1), transcript variant 2, mRNA | NM_001159920.1 | NP_001153392.1 | <1 |
| GAB1 | GRB2-associated binding protein 1 (GAB1), transcript variant 1, mRNA | NM_207123.2 | NP_997006.1 | <1 |
| GGN | Gametogenetin (GGN), mRNA | NM_152657.3 | NP_689870.3 | <1 |
| GPA33 | Glycoprotein A33 (transmembrane) (GPA33), mRNA | NM_005814.1 | NP_005805.1 | <1 |
| GTF2IRD1 | GTF2I repeat domain containing 1, mRNA (cDNA clone MGC:9316 IMAGE:3913745), complete CDS | BC018136.1 | AAH18136.1 | <1 |
| H2AFX | H2A histone family, member X (H2AFX), mRNA | NM_002105.2 | NP_002096.1 | <1 |
| HIF1A | Hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) (HIF1A), transcript variant 2, mRNA | NM_181054.2 | NP_851397.1 | <1 |
| HK2 | Hexokinase 2 (HK2), mRNA | NM_000189.4 | NP_000180.2 | <1 |
| IGF1 | Insulin-like growth factor 1 (somatomedin C) (IGF1), transcript variant 4, mRNA | NM_000618.3 | NP_000609.1 | <1 |
| IGF1R | Insulin-like growth factor 1 receptor (IGF1R), mRNA | NM_000875.3 | NP_000866.1 | <1 |
| IGFBP4 | Insulin-like growth factor binding protein 4 (IGFBP4), mRNA | NM_001552.2 | NP_001543.2 | <1 |
| KRT14 | Keratin 14 (KRT14), mRNA | NM_000526.4 | NP_000517.2 | <1 |
| LARGE | Glycosyltransferase-like protein (LARGE), transcript variant 1, mRNA | NM_004737.4 | NP_004728.1 | <1 |
| LIN28A | Lin-28 homolog A (C. elegans) (LIN28A), mRNA | NM_024674.4 | NP_078950.1 | <1 |
| MAP2K7 | Mitogen-activated protein kinase kinase 7, mRNA (cDNA clone MGC:33283 IMAGE:5269063), complete CDS | BC038295.2 | AAH38295.1 | <1 |
| MAS1 | MAS1 oncogene (MAS1), mRNA | NM_002377.2 | NP_002368.1 | <1 |
| MDM2 | Mdm2, p53 E3 ubiquitin protein ligase homolog (mouse) | Q00987 | Q00987 | <1 |
| MLANA | Melan-A (MLANA), mRNA | NM_005511.1 | NP_005502.1 | <1 |
| MPL | Myeloproliferative leukemia virus oncogene (MPL), mRNA | NM_005373.2 | NP_005364.1 | <1 |
| NMB | Neuromedin B, mRNA (cDNA clone MGC:17211 IMAGE:4185578), complete CDS | BC008603.1 | AAH08603.1 | <1 |
| PIK3CA | Phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha (PIK3CA), mRNA | NM_006218.2 | NP_006209.2 | <1 |
| PTGER1 | Prostaglandin E receptor 1 (subtype EP1), 42 kDa (PTGER1), mRNA | NM_000955.2 | NP_000946.2 | <1 |
| SFTPB | Surfactant protein B, mRNA (cDNA clone MGC:45314 IMAGE:5179408), complete CDS | BC032785.1 | AAH32785.1 | <1 |
| SLAMF7 | SLAM family member 7, mRNA (cDNA clone MGC:34188 IMAGE:5203638), complete CDS | BC027867.1 | AAH27867.1 | <1 |
| SOX2 | SRY (sex determining region Y)-box 2 (SOX2), mRNA | NM_003106.3 | NP_003097.1 | <1 |
| SRC | V-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) (SRC), transcript variant 1, mRNA | NM_005417.3 | NP_005408.1 | <1 |
| TSG101 | Tumor susceptibility gene 101, mRNA (cDNA clone MGC:1317 IMAGE:3350039), complete CDS | BC002487.1 | AAH02487.1 | <1 |
| WT1 | Wilms tumor 1, mRNA (cDNA clone MGC:33649 IMAGE:4827849), complete CDS | BC032861.2 | AAH32861.1 | <1 |
| XRCC1 | X-ray repair complementing defective repair in Chinese hamster cells 1 (XRCC1), mRNA | NM_006297.2 | NP_006288.2 | <1 |
| YBX1 | Y box binding protein 1 (YBX1), mRNA | NM_004559.3 | NP_004550.2 | <1 |
For Research Use Only. Not for use in diagnostic procedures.