Search
The septum you choose for your autosampler vial can impact sample evaporation, sample security, needle bending and blockages, and instrument down time. Don’t let the complexities of septa selection slow you down. Use the simple online Thermo Scientific SureSTART Selection Guide to find the right septa and vial for your analysis.
Answer a few simple questions and our SureSTART Selection Guide will recommend the best septa, cap, and vial for your workflow. You can also enter your current septa, cap, and vial part numbers into our equivalence tool to identify equivalent or better-performing Thermo Scientific SureSTART products.
The Septa Selection Tool makes use of the chart below to select a septum that will help prevent contamination and avoid damage.
| Multiple injection? | Temperature? | Thin, fragile needle? | Blunt, thick needle? | Critical analysis? | Low coring? | ||
|---|---|---|---|---|---|---|---|
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ||
| Good resealability required | -40°C up to 120°C | -40°C up to 110°C | -60°C up to 200°C | Soft and thin septa required | Slit/pre-cut liner as penetration aid (HPLC) | Very clean liner required | Double-sided PTFE laminated liners required |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| Natural rubber/TEF | Natural rubber/TEF; Butyl/PTFE | Red rubber/PTFE | Silicone/PTFE | e.g. Silicone/PTFE | Silicon/PTFE, slit | Silicon/PTFE | PTFE/Silicone/PTFE PTFE/Butyl/PTFE |
When selecting which septa is right for your experiments, it is important to consider the:
Each instrument vendor uses different injector needle types and for some instruments, having pre-slit septa, or choosing a material with a lower shore (hardness) value will help reduce the chances of needle issues, such as needle bending.
If your compounds are volatile then choosing septa that will provide a tight seal is key to preventing compound loss. Our Advanced Vial Closure System (AVCS) caps remove subjectivity around achieving the optimal seal compression when closing a vial.
Some solvents that are commonly used for HPLC, UHPLC and GC experiments can impact the septa integrity.
If you need to place a capped sample vial in a high temperature environment, then you will need to consider septa materials that are able to withstand high temperatures.
Septa are made from different materials. See below for definitions and uses of different types of septa materials.
Used primarily for routine analysis in gas chromatography. Offers moderate ability to reseal and good chemical inertness. Not recommended for multiple injections or holding samples for further analysis. PTFE is protective layer that once broken exposes rubber to chemical attack.
Low durometer of rubber allows ease of needle penetration. A popular and economical septum for general GC purposes.
Harder grade of rubber for use with piercing needle. Most popular and economical septa for general GC purposes.
Pre-slit, high quality red rubber with a thin (0.003") layer PTFE. For applications using a very thin-gauge syringe needle or in instances when a vacuum may form in the vial.
High quality, silicone rubber laminated to PTFE. Use when excellent resealing qualities are a must. Septum resists coring and is recommended when multiple injections are required. Preferred septa for use in liquid chromatography applications.
A white medium hardness silicone with red PTFE protective layer available in a range of thickness.
A very pure soft silicone laminated to PTFE. Septum resists coring and is recommended for instruments with fine gauge needles. Also recommended for LC-MS and GC-MS due to high purity.
A very soft silicone laminated to PTFE. Use with flexible needle.
A layer of PTFE on each side of medium hardness silicone. Most resistant to coring with above average resealing characteristics. Recommended for most demanding applications such as trace analysis, longer time between injections, or for internal standards. Use with any autosampler using large diameter, blunt-tip syringe needles.
Pre-slit, high quality pure white silicone faced with PTFE. For applications using a very thin-gauge syringe needle or in instances when a vacuum may form in the vial.
Very good chemical resistance and used as a protective layer for less resistant elastomers.
For single injections and short sample cycles. This type of septa is not resealable.
Viton provides the best chemical resistance with limited ability to reseal. Recommended for chlorinated solvents. Due to Viton’s intrinsic hardness, these septa are not suitable for finer-gauge syringe needles.
Moulded as part of the cap.
Chemically resistant but for one time use only with no ability to reseal. Free of fluoropolymer coating, so suitable for PFOA analysis. 20 mm seal selection for headspace and sample preparation applications.
An economical choice for low temperature (< 125°C) or low-pressure applications. Not suitable for alkanes, benzene, chlorinated solvents or cyclohexane without a protective PTFE layer.
Does not provide PTFE barrier. Use for gas sampling due to low permeability.
Does not provide PTFE barrier. Use for gas sampling due to low permeability.
Has PTFE barrier that makes it suitable for work with general organic solvents with low gas permeability.
A specially moulded seal with a PTFE insert. Sealing surface of Butyl and PTFE affects a more positive seal than non-PTFE-faced septa. Ideal choice for temperatures below 125°C. Good sealing characteristics, excellent resistance to most solvents with reduced coring and high puncture tolerance. PTFE provides increased chemical resistance.
Excellent septa choice for volatiles with very low background peaks and low permeability. Also ideal for alcohols and aqueous samples. Good resealing characteristics and resistant to coring.
Best septa choice when temperatures are over 125°C.
High temperature formulated seal with low bleed. Best septa choice when temperatures are up to 300°C.
Thin 1.4 mm seal with PTFE face. Resealing capability limited due to thinner silicone layer.
Reflective aluminum face protects the silicone seal. The white silicone is suitable for use up to 170°C.
Reflective aluminum face protects the silicone seal. The red silicone is suitable for use at temperatures up to 250°C.
Soft silicone with clean formulation for minimal interference. Thinner seal suitable for solvent washing, solvent extraction, and solid phase microextraction (SPME) applications with some resealing. Not for direct headspace applications.
Butyl bungs for sealing of lyophilized products. Compatible with low storage temperatures and low gas permeability.
Thin PTFE layer with sealing ring to give secure closure for strong solvents. For use in liquid extraction or SPME stage during sample preparation. Does not reseal. Single use only.
Use Thermo Scientific 2 mL SureStop Vials with Advanced Vial Closure System (AVCS) Caps
Innovative screw cap & vial design ensures proper screw cap seal every time, independent of operator, so there’s no more failures from:
Now you can reduce re-analysis costs and increase data integrity and sample throughput.