Accelerate and simplify your workflows with the KingFisher SpeciTRAX Sample Transfer System. This innovative system combines new sample-handling automation technologies to offer ease of use, as well as increased throughput and safety, while delivering consistent, accurate results.
The KingFisher SpeciTRAX system enables your lab to transfer 192 samples from capped sample tubes to two 96 deep-well plates in 40 minutes, all within a closed-door system.
Virtually navigate the SpeciTRAX instrument, get a closer view of features, and click to learn more about specific components.
Note: This tool is best viewed in Chrome, Firefox, or Safari browser. Learn more
Minimize risk with safety with built-in features
Increase consistency and accuracy with sample tracking and precision pipettes
Adoption is easy with surefire design and intuitive software
Customize workflows
An open software platform allows for easy-to-make changes in the user interface (UI) or an Excel file for customized applications or workflow modifications. Parameter variations for tubes, plates, aspiration, and dispensing volumes for specific protocols are easily accommodated. Liquid handling parameters and transfer volumes are simply configured or preset for a specific protocol. Plate layout and sample transfer paths can also be predefined; this option is convenient for situations when specific wells need to be blank for manual transfers such as controls.
Model |
KingFisher SpeciTRAX Sample Transfer System | |
Sizing information |
Instrument dimensions |
7 ft (L) x 5.5 ft (W) 2.1 m (L) x 1.5 m (W) |
Clearance space |
7.1 ft (H) 2.1 m (H) |
|
Weight |
1200 lbs 550 kg |
|
Electrical requirements |
Power |
120/240V |
Hz |
50/60 Hz |
|
Power consumption |
1000 VA |
|
Operating environment |
Temperature |
19–25°C (66.2–77°F) |
Relative humidity |
20–80% RH, non-condensing |
|
Atmospheric pressure |
80–106 kPa |
|
Functions |
Integrate multiple functions, including decapping, recapping, barcode identification, automated liquid transfer, HEPA-filtration and negative pressure protection |
|
Throughput |
192 samples/40 min | |
Pipetting precision |
Pipetting method |
Based on the principle of air displacement, liquid level detection |
Volume range |
10–1,000 µL |
|
Precision (%CV) |
10 μL: ≤ 0.1% 100 μL: ≤ 0.2% 1,000 μL: ≤ 2.5% |
|
Accuracy (%DEV) |
10 μL: ≤ ±1.7% 100 μL: ≤ ±2.9% 1,000 μL: ≤ ±9.9% |
|
Robot arm accuracy |
x, y, z: ±0.1 mm | |
Barcode scanner |
4 scanner sensors, supports both 1D and 2D barcodes |
|
Protection | UV light |
36 W |
Supply and exhaust air filter |
ISO 5, HEPA 99.995% at 0.3 µm, negative pressure system |
|
Tube compatibility |
Universal fit. Excellent fit on 5 mL and 10 mL transport tubes with screw caps. |
For Research Use Only. Not for use in diagnostic procedures.