What are special handling options for oligo orders?
When you check Special Handling on any of the oligo order pages, the page is updated with the special handling selections. You can select from the following special handling options:
  1. Extra Label: an extra copy of the tube label is sent for record keeping or labeling additional tubes.
  2. 100% QC: ensures the oligo is QC tested by mass spectrometry. Applies only to 25N and 50N DSL oligos. Mass spec traces are not sent out. Oligos that are modified, purified, or ≥200N scale go through this procedure by default.
  3. Standard Concentration: normalization of all oligos to a set concentration in water.