Search
Our base systems support a range of pressures, flow rates, column dimensions, and detector options to meet your application and performance needs.
Vanquish Core HPLC Systems![]() | Vanquish Flex UHPLC Systems![]() | Vanquish Horizon UHPLC Systems![]() | Vanquish Neo UHPLC System![]() |
| Download brochure | Download brochure | Download brochure | Download brochure |
| Description | |||
|---|---|---|---|
| A dependable workhorse for your high-volume QC analysis | Run HPLC and UHPLC analyses on a single system | Run smaller particles and column dimensions with confidence | Delivers robust separations 24/7 to support your high-sensitivity LC-MS workflows |
| Pump options | |||
| Binary, quaternary, dual & isocratic | Binary, quaternary & dual | Binary | Binary |
| Maximum system pressure | |||
| 700 bar | 1000 bar | 1500 bar | 1500 bar |
| Flow range | |||
| 0.001–10 mL/min | 0.001–8 mL/min | 0.001–5 mL/min | 1 nL/min–100 μL/min |
| Column particle size | |||
| 3–5 µm | 1.5–3 µm | < 3 µm | ≤ 3 µm |
| Shop now | Shop now | Shop now | Shop now |
| My Green Lab® ACT® Ecolabel | |||
| See label | See label | See label | See label |
Our application-specific instruments can help you maximize productivity, sample knowledge and purity, or optimize methods.
| Vanquish Amplify UHPLC Systems ![]() | Vanquish Duo HPLC & UHPLC Systems![]() | Vanquish Online 2D-LC Systems ![]() | Vanquish Analytical Purification LC Systems![]() | Vanquish Method Development System![]() |
| System feature | ||||
|---|---|---|---|---|
| Improve separations of metal-sensitive molecules by minimizing adsorption and reducing metal adduct formation | Run two analyses, simultaneously, on one instrument | Improve analysis of complex samples or simplify separation of poorly resolved analytes | High-performance fractionation capabilities to deliver you ultra-pure products | Accelerate and optimize your method by testing different buffers mixtures across six columns |
| Shop now | Shop now | Shop now | Shop now | Shop now |
Featuring a handful of new enhancements with replaceable parts to help increase your return on investment and support your sustainability goals. Our Vanquish LC Systems are now easier to use, more reliable, and include torque-controlled fingertight fittings as standard.
Easier to use
Faster workflows
More sustainable parts
Discover all of our My Green Lab ACT Ecolabel chromatography products
Using the new fluidic connections with Viper TQ UHPLC Fitting design on our Vanquish LC systems has been a great experience. The robust and tight connections provide a high level of reliability, and the capillaries are easier to use compared to other solutions. The tactile feedback ensures that the fittings are properly secured, regardless of the columns used. I also value the active pre-heater TQ design with its replaceable outlet capillaries, which improves ecological efficiency while providing performance and consistency in my analyses. The addition of the new eluent filters is another great benefit, making the overall system even more user-friendly.
— Lydia D. | Pharma
I love my Vanquish HPLC so much because its performance is so reliable. During the pandemic, we didn't get a chance to work on it, and it was turned off for over a year. But after we returned to the lab, we did quick routine maintenance and injected some QC samples. The retention times of the standards aligned SO WELL with previous QCs. I was absolutely in awe of the reproducibility. I would definitely recommend Vanquish to my colleagues.
— Harong L. | Academia
My Vanquish UHPLC, named Cynthia, has been a fantastic addition to my lab space. Fast run times, robust methodology, and the ease of use for Chromeleon makes Cynthia a vital part of my lab team!
— Bjorn L. | Industry
We performed a method translation from a dated HPLC method to UHPLC on the Vanquish. The new method allows this test to be performed within one shift instead of across multiple shifts/personnel.
— Nicholas G. | Pharma
* Required field