pSilencer™ 3.1-H1 puro
AM5768: pSilencer™ 3.1-H1 puro
PROPRIETARY -- DO NOT DISCLOSE
Features:
NOTE: This sequence is for the new and improved p Silencer 3.1-H1 puro vector which began shipping in Jan 2004.
Please see the New Vector Map for diagram, annotated sequence, and restriction sites of our new vector.
Vector Size: 4121 bp
Promoter: Human H1
Ampicillin resistance gene
Puromycin resistance gene (SV40 early promoter, SV40 early polyadenylation signal)
ColE1 origin of replication
PROPRIETARY -- DO NOT DISCLOSE
Features:
NOTE: This sequence is for the new and improved p Silencer 3.1-H1 puro vector which began shipping in Jan 2004.
Please see the New Vector Map for diagram, annotated sequence, and restriction sites of our new vector.
Vector Size: 4121 bp
Promoter: Human H1
Ampicillin resistance gene
Puromycin resistance gene (SV40 early promoter, SV40 early polyadenylation signal)
ColE1 origin of replication
We recommend the following primers for sequencing:
M13F(-40): 5'-GTTTTCCCAGTCACGAC-3'
3.0rev: 5'-GAGTTAGCTCACTCATTAGGC-3'
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (66 base pairs) between the BamH I and Hind III sites.
M13F(-40): 5'-GTTTTCCCAGTCACGAC-3'
3.0rev: 5'-GAGTTAGCTCACTCATTAGGC-3'
Note: This is the sequence of the negative control vector provided with the kit. It contains a negative control siRNA template (66 base pairs) between the BamH I and Hind III sites.
Sequence:
For Research Use Only. Not for use in diagnostic procedures.