Search
Isotype controls should be matched to the primary antibody’s host species and class, including light chains (e.g., IgG2 Lambda, IgA1 Kappa). If using directly labeled primary antibodies, it is important to use the same conjugate to avoid differences in detection.
Although isotype controls are mainly used in flow cytometry, they can be used as standard blocking agents and protein coating agents for other applications including immunofluorescence, immunocytochemistry, western blotting, and ELISA.
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
For Research Use Only. Not for use in diagnostic procedures.